Skip to Content
Merck
All Photos(5)

Key Documents

212857

Sigma-Aldrich

Rhodium on alumina

greener alternative

extent of labeling: 5 wt. % loading, matrix alumina support, powder

Synonym(s):

Rh/Al2O3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: rhodium
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

5 wt. % loading

matrix

alumina support

greener alternative category

SMILES string

[Rh]

InChI

1S/Rh

InChI key

MHOVAHRLVXNVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Rhodium on alumina (Rh/Al2O3) (5 wt. % loading, matrix alumina support, powder) is utilized as a catalyst for the steam reforming of ethanol for use in hydrogen production using molten carbonate fuel cells. Rh/Al2O3 can also be used as a catalyst in the production of hydrogen from sodium borohydride (NaBH4).

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calorimetric study on oxidation of alumina supported rhodium by dioxygen
Wang, CB, et al.
J. Mol. Catal. A: Chem., 120(1), 179-184 (1997)
Real-time XAFS analysis of Rh/alumina catalyst
Dohmae K, et al.
Surface and Interface Analysis : SIA, 40(13), 1751-1754 (2008)
Performance of Rh/Al2O3 catalyst in the steam reforming of ethanol: H2 production for MCFC
Cavallaro S, et al.
Applied Catalysis A: General, 249(1), 119-128 (2003)
Comparative XPS study of Rh/Al2O3 and Rh/TiO2 as catalysts for NaBH4 hydrolysis
Larichev YV, et al.
International Journal of Hydrogen Energy, 35(13), 6501-6507 (2010)
Ethanol steam reforming on Rh/Al2O3 catalysts
Cavallaro S
Energy & Fuels, 14(6), 1195-1199 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service