Skip to Content
Merck
All Photos(2)

Documents

206172

Sigma-Aldrich

Rhodium on alumina

greener alternative

extent of labeling: 0.5 wt. % loading, pellets, 3.2 mm

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

pellets

Quality Level

reaction suitability

core: rhodium
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

0.5 wt. % loading

particle size

3.2 mm

greener alternative category

SMILES string

[Rh]

InChI

1S/Rh

InChI key

MHOVAHRLVXNVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Rhodium on alumina (Rh/Al2O3) (0.5 wt.% loading, pellets, 3.2 mm) is utilized as a catalyst for the steam reforming of ethanol for use in hydrogen production using molten carbonate fuel cells. It can also be used as a catalyst in the production of hydrogen from sodium borohydride (NaBH4).

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance of Rh/Al2O3 catalyst in the steam reforming of ethanol: H2 production for MCFC
Cavallaro S, et al.
Applied Catalysis A: General, 249(1), 119-128 (2003)
Ethanol steam reforming on Rh/Al2O3 catalysts
Cavallaro S
Energy & Fuels, 14(6), 1195-1199 (2000)
Comparative XPS study of Rh/Al2O3 and Rh/TiO2 as catalysts for NaBH4 hydrolysis
Larichev YV, et al.
International Journal of Hydrogen Energy, 35(13), 6501-6507 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service