Skip to Content
Merck
All Photos(3)

Documents

205710

Sigma-Aldrich

Palladium on alumina

greener alternative

extent of labeling: 5 wt. % loading, powder

Synonym(s):

Pd

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

5 wt. % loading

mp

1552 °C (lit.)

greener alternative category

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Palladium on alumina (Pd/Al2O3) (5 wt % loading, powder) is a widely utilized catalyst that can be prepared by wet impregnation of alumina pellets with palladium chloride. It can also be prepared by flame spray pyrolysis (FSP) by utilizing the precursor solution of aluminum sec-butoxide and palladium acetylacetonate.

Application

Pd/Al2O3 can be used as a catalyst in the enantioselective hydrogenation reaction. It can also be potentially used in the formation of an automotive catalytic converter to improve the air quality of urban environments.

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aqueous bromate reduction by catalytic hydrogenation over Pd/Al2O3 catalysts
Chen H, et al.
Applied Catalysis. B, Environmental, 96(3-4), 307-313 (2010)
Catalytic activity of Pd/Al2O3 toward the combustion of methane
Gao D, et al.
Catalysis Communications, 9(15), 2583-2587 (2008)
Identification of the phytosphingosine metabolic pathway leading to odd-numbered fatty acids
Kondo N, et al.
Nature Communications, 5(1), 1-13 (2014)
Flame spray synthesis of Pd/Al2O3 catalysts and their behavior in enantioselective hydrogenation
Strobel R, et al.
J. Catal., 222(2), 307-314 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service