Skip to Content
Merck
All Photos(2)

Key Documents

205583

Sigma-Aldrich

Tetraethylammonium acetate tetrahydrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(OCOCH3) · 4H2O
CAS Number:
Molecular Weight:
261.36
Beilstein:
3574285
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

42-46 °C (lit.)

solubility

water: 100 mg/mL, clear, colorless

functional group

amine

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC([O-])=O.CC[N+](CC)(CC)CC

InChI

1S/C8H20N.C2H4O2.4H2O/c1-5-9(6-2,7-3)8-4;1-2(3)4;;;;/h5-8H2,1-4H3;1H3,(H,3,4);4*1H2/q+1;;;;;/p-1

InChI key

QNOAKQGNSWGYOE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tetraethylammonium acetate tetrahydrate was used in the preparation of transport membrane which selectively permeates carbon dioxide from hydrogen and methane. It was employed in the ring opening of 1,3,2-dioxaphospholanes and in the synthesis of ether phospholipids.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Chemical Communications, 70-70 (1993)
New facilitated transport membranes for the separation of carbon dioxide from hydrogen and methane.
Quinn R, et al.
Journal of Membrane Science, 104(1), 139-146 (1995)
Tetrahedron Letters, 33, 2291-2291 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service