Skip to Content
Merck
All Photos(2)

Key Documents

195731

Sigma-Aldrich

Iron(0) pentacarbonyl

greener alternative

Synonym(s):

Iron(0) carbonyl, Pentacarbonyliron(0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(CO)5
CAS Number:
Molecular Weight:
195.90
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.74 (vs air)

Quality Level

vapor pressure

35 mmHg ( 25 °C)
40 mmHg ( 30.3 °C)

autoignition temp.

122 °F

reaction suitability

core: iron
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.5196 (lit.)

bp

103 °C (lit.)

mp

−20 °C (lit.)

density

1.49 g/mL at 25 °C (lit.)

greener alternative category

storage temp.

2-8°C

SMILES string

[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/5CO.Fe/c5*1-2;

InChI key

FYOFOKCECDGJBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Iron(0) pentacarbonyl can be used:
  • As an iron source to synthesize Fe-doped thin TiO2 films.
  • As a ligand to synthesize complexes of gold(I) and Fe(CO)5, silver and Fe(CO)5.
Reactant for:
  • Addition reactions
  • Decomposition via photolysis
  • Preparation of magnetic Fe onion-like fullerene micrometer-sized particles of narrow size distribution via thermal decomposition
  • Preparation of a heptanuclear iron carbonyl cluster used in catalytic hydrosilane reduction of carboxamides

  • Precursor for preparation of Fe nanoparticles for photocatalytic hydrogen evolution under highly basic conditions

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Djordje Miljković et al.
Journal of neurochemistry, 118(2), 205-214 (2011-05-11)
Experimental autoimmune encephalomyelitis (EAE) is a model of multiple sclerosis. Dark Agouti rats immunized with spinal cord homogenate (SCH) and carbonyl iron (CI), as an adjuvant, develop severe hyperacute form of EAE. They succumb to EAE earlier and have higher
Tetsu Mitsumata et al.
The journal of physical chemistry. B, 116(40), 12341-12348 (2012-09-15)
A new class of magnetoelastic gel that demonstrates drastic and reversible changes in storage modulus without using strong magnetic fields was obtained. The magnetic gel consists of carrageenan and carbonyl iron particles. The magnetic gel with a volume fraction of
Lindsay Hewison et al.
Dalton transactions (Cambridge, England : 2003), 39(38), 8967-8975 (2010-08-18)
New CO-releasing molecules, CO-RMs, based on indenyl iron carbonyls have been identified. Half-lives for carbon monoxide (CO) release, (1)H NMR, (13)C NMR, IR, mass spectra, elemental analysis and biological data have been determined for the compounds. Limited correlations have been
Heterobimetallic Complexes Featuring Fe(CO)5 as a Ligand on Gold.
Wang G, et al.
Chemistry?A European Journal , 23(68), 17222-17226 (2017)
Plasma enhanced chemical vapor deposition of iron doped thin dioxide films, their structure and photowetting effect.
Sobczyk-Guzenda A, et al.
Thin Solid Films, 589, 605-612 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service