Skip to Content
Merck
All Photos(2)

Documents

241431

Sigma-Aldrich

Tungsten hexacarbonyl

97%

Synonym(s):

Hexacarbonyltungsten, Tungsten carbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
W(CO)6
CAS Number:
Molecular Weight:
351.90
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

12.1 (vs air)

vapor pressure

1.2 mmHg ( 67 °C)

Assay

97%

form

solid

reaction suitability

core: tungsten
reagent type: catalyst

impurities

<1% Mo

mp

150 °C (lit.)

density

2.65 g/mL at 25 °C (lit.)

SMILES string

[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/6CO.W/c6*1-2;

InChI key

FQNHWXHRAUXLFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to a Fischer carbene complex which reduces Pd(II) to nanoparticulate Pd(0) that is catalytically active in Hiyama cross-coupling.
Tungsten hexacarbonyl, [W(CO)6] may be used as a precursor for the deposition of WO3-x films, which may be used as gas sensors for the detection of NO2.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dipankar Srimani et al.
Organic letters, 9(18), 3639-3642 (2007-08-07)
An efficient synthesis of Pd nanoparticles in water has been developed using a Fischer carbene complex of tungsten as the reductant and PEG as the capping agent. The colloidal palladium (1 mol %) efficiently catalyzes Hiyama cross-coupling reactions performed in
Aerosol assisted chemical vapour deposition of WO 3 thin films from tungsten hexacarbonyl and their gas sensing properties.
Ashraf S, et al.
Journal of Materials Chemistry, 17.35, 3708-3713 (2007)
Keisha-Gay Hylton et al.
The Journal of organic chemistry, 68(4), 1615-1617 (2003-02-15)
W(CO)(6)-catalyzed carbonylation provides an alternative to phosgene or phosgene derivatives such as 1,1-carbonyldiimidazole (CDI) for the conversion of amines to ureas. As an illustration, the core structure of the HIV protease inhibitors DMP 323 and DMP 450 has been prepared
Seth M Dumbris et al.
The Journal of organic chemistry, 74(22), 8862-8865 (2009-10-30)
Hydantoins can be synthesized from the corresponding amino amides employing oxidative catalytic carbonylation using W(CO)(6) as the catalyst, I(2) as the oxidant, CO as the carbonyl source, and DBU as base. Secondary amides afford the hydantoins in good to excellent
S Martin et al.
The Journal of chemical physics, 134(3), 034304-034304 (2011-01-26)
We have studied the excitation and dissociation processes of the molecule W(CO)(6) in collisions with low kinetic energy (3 keV) protons, monocharged fluorine, and chlorine ions using double charge transfer spectroscopy. By analyzing the kinetic energy loss of the projectile

Articles

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service