Skip to Content
Merck
All Photos(1)

Documents

213748

Sigma-Aldrich

1,2-Hexadecanediol

technical grade, 90%

Synonym(s):

1,2-Dihydroxyhexadecane, 1,2-Hexadecylene glycol, 2-Hydroxycetyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CHOHCH2OH
CAS Number:
Molecular Weight:
258.44
Beilstein:
1722206
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

mp

68-72 °C (lit.)

SMILES string

CCCCCCCCCCCCCCC(O)CO

InChI

1S/C16H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16(18)15-17/h16-18H,2-15H2,1H3

InChI key

BTOOAFQCTJZDRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Hexadecanediol was used as reducing agent in the preparation of silver nanocrystals from a dichlorobenzene solution containing oleyl amine as a surfactant. It was also used in the preparation of:
  • Au-Fe3O4 hetero-dimers
  • iron pyrite nanocrystals
  • monodisperse iron platinum nanoparticles

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

361.4 °F - closed cup

Flash Point(C)

183 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leize Zhu et al.
Nanoscale, 6(2), 1029-1037 (2013-12-03)
Earth-abundant and nontoxic pyrite iron disulfide (FeS2) is very promising for photovoltaic applications but the phase purity and the morphology of iron pyrite nanocrystals (NCs) have a significant impact on the solar cell performance. In this work, we systematically investigated
Yongmin Ko et al.
Nature communications, 8(1), 536-536 (2017-09-16)
The effective implantation of conductive and charge storage materials into flexible frames has been strongly demanded for the development of flexible supercapacitors. Here, we introduce metallic cellulose paper-based supercapacitor electrodes with excellent energy storage performance by minimizing the contact resistance
Deepak B Thimiri Govinda Raj et al.
Molecular systems biology, 7, 541-541 (2011-10-27)
We manufactured a novel type of lipid-coated superparamagnetic nanoparticles that allow for a rapid isolation of plasma membranes (PMs), enabling high-resolution proteomic, glycomic and lipidomic analyses of the cell surface. We used this technology to characterize the effects of presenilin
Yang Sheng et al.
Journal of colloid and interface science, 374(1), 96-101 (2012-03-01)
Au-Fe(3)O(4) composite nanoparticles have received much research interest due to their promising biomedical applications. In this work, Au-Fe(3)O(4) composites with well-defined dimer-like nanostructure were synthesized via thermal decomposition route. The surfactant 1,2-hexandicandiol has proved to be critical for the formation
Tobias Preller et al.
Nanomaterials (Basel, Switzerland), 8(5) (2018-05-13)
The synthesis of FePt nanocrystals is typically performed in an organic solvent at rather high temperatures, demanding the addition of the in situ stabilizers oleic acid and oleylamine to produce monomodal particles with well-defined morphologies. Replacing frequently-used solvents with organic

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service