Skip to Content
Merck
All Photos(1)

Key Documents

185086

Sigma-Aldrich

Sodium cyanate

96%

Synonym(s):

Sodium isocyanate, Zassol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOCN
CAS Number:
Molecular Weight:
65.01
Beilstein:
3655041
EC Number:
MDL number:
UNSPSC Code:
12161600
eCl@ss:
38060512
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

reaction suitability

core: sodium
reagent type: catalyst

SMILES string

[Na+].[O-]C#N

InChI

1S/CHNO.Na/c2-1-3;/h3H;/q;+1/p-1

InChI key

ZVCDLGYNFYZZOK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rivera-Ch et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 164(8), 659-662 (1995-01-01)
The Hb-O2 affinity and the erythropoietic response as a function of time were studied in mice treated with sodium cyanate for up to 2 months. Cyanate increased the Hb-O2 affinity in normoxic mice more than in chronically hypoxic mice. The
Louis M P Ter-Ovanessian et al.
Scientific reports, 11(1), 19356-19356 (2021-10-01)
The first step of pyrimidine synthesis along the orotate pathway is studied to test the hypothesis of geochemical continuity of protometabolic pathways at the origins of life. Carbamoyl phosphate (CP) is the first high-energy building block that intervenes in the
J Tor-Agbidye et al.
Brain research, 820(1-2), 12-19 (1999-02-19)
Sodium cyanate, a neurotoxic chemical in rodents, primates and humans, is implicated in neurodegenerative disorders in protein-deficient populations subsisting in parts of Africa on the cyanogenic plant cassava. The molecular and cellular mechanisms of cyanate neurotoxicity are not understood. This
Yuki Taga et al.
Cell chemical biology, 24(10), 1276-1284 (2017-09-26)
Carbamylation is a non-enzymatic post-translational modification that physiologically occurs during aging and is a risk factor for various diseases. The most common product of carbamylation is homocitrulline (HCit), where a lysine (Lys) amino group has reacted with urea-derived cyanate. HCit has recently been detected
K K Henderson et al.
Journal of applied physiology (Bethesda, Md. : 1985), 89(1), 265-272 (2000-07-25)
These studies were conducted to compare the effects on systemic O(2) transport of chronically vs. acutely increased Hb O(2) affinity. O(2) transport during maximal normoxic and hypoxic [inspired PO(2) (PI(O(2))) = 70 and 55 Torr, respectively] exercise was studied in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service