Skip to Content
Merck
All Photos(1)

Key Documents

1.06437

Supelco

Sodium cyanide

EMPLURA®

Synonym(s):

Sodium cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CNNa
CAS Number:
Molecular Weight:
49.01
MDL number:
UNSPSC Code:
12352302
EC Index Number:
205-599-4
NACRES:
NA.21
Assay:
≥95.0% (silver nitrate titration)
form:
solid
solubility:
370 g/L

Quality Level

product line

EMPLURA®

Assay

≥95.0% (silver nitrate titration)

form

solid

potency

4.7 mg/kg LD50, oral (Rat)
7.7 mg/kg LD50, skin (Rabbit)

pH

11.7 (20 °C, 100 g/L in H2O)

bp

1496 °C/1013 hPa

mp

563 °C

solubility

370 g/L

density

1.6 g/cm3 at 20 °C

bulk density

750‑900 kg/m3

anion traces

chloride (Cl-): ≤0.05%
hexacyanoferrate(II) ([Fe(CN)6]4-): ≤0.05%
sulfide (S2-): ≤0.02%

cation traces

Pb: ≤0.005%

storage temp.

no temp limit

SMILES string

[Na+].N#[C-]

InChI

1S/CN.Na/c1-2;/q-1;+1

InChI key

MNWBNISUBARLIT-UHFFFAOYSA-N

Application


  • Green Synthesis of Sodium Cyanide Using Hydrogen Cyanide Extracted under Vacuum from Cassava (Manihot esculenta Crantz) Leaves.: This study presents an innovative green approach to synthesizing sodium cyanide using hydrogen cyanide extracted from cassava leaves. The process involves maceration and vacuum extraction, producing NaCN with potential applications in environmentally friendly chemical synthesis and analytical methods (Monga et al., 2022).

Analysis Note

Assay (argentometric): ≥ 95.0 %
Identity: passes test
Chloride (Cl): ≤ 0.05 %
Hexacyanoferrate: ≤ 0.05 %
Sulphide (S): ≤ 0.02 %
Pb (Lead): ≤ 0.005 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service