Skip to Content
Merck
All Photos(3)

Key Documents

178470

Sigma-Aldrich

4-(Trifluoromethyl)phenol

97%

Synonym(s):

α,α,α-Trifluoro-p-cresol, 4-Hydroxybenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4OH
CAS Number:
Molecular Weight:
162.11
Beilstein:
1637019
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

mp

45-47 °C (lit.)

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)C(F)(F)F

InChI

1S/C7H5F3O/c8-7(9,10)5-1-3-6(11)4-2-5/h1-4,11H

InChI key

BAYGVMXZJBFEMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-(Trifluoromethyl)phenol molecule, bound at the active site of H61T (His-61→Thr) mutant, shows strong density.

4-(Trifluoromethyl)phenol also known as p-trifluoromethylphenol, is used in synthesis of polymers and monomers.

Application

4-(Trifluoromethyl)phenol (4-hydroxybenzotrifluoride) was used in the synthesis of diaryl ether.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jin-Kyun Lee et al.
Chemical communications (Cambridge, England), (39)(39), 4780-4782 (2008-10-03)
A high yielding, batch mode synthesis of diaryl ethers and sulfides by an S(N)Ar fluoride-mediated process in scCO(2) has been developed; the use of a polymer-supported imidazolium fluoride reagent in batch mode led to the development of a fixed-bed continuous
B van de Wier et al.
Chemico-biological interactions, 242, 139-144 (2015-10-03)
Cytochrome P450 2E1 (CYP2E1) expression and activity in the liver is associated with the degree of liver damage in patients with alcoholic steatohepatitis (ASH) as well as non-alcoholic steatohepatitis (NASH). CYP2E1 is known to generate reactive oxygen species, which leads
M W Fraaije et al.
The Journal of biological chemistry, 275(49), 38654-38658 (2000-09-14)
Vanillyl-alcohol oxidase (VAO) is member of a newly recognized flavoprotein family of structurally related oxidoreductases. The enzyme contains a covalently linked FAD cofactor. To study the mechanism of flavinylation we have created a design point mutation (His-61 --> Thr). In
Selina Tisler et al.
Environmental science & technology, 53(13), 7400-7409 (2019-05-29)
The present study investigates the transformation of the antidepressant fluoxetine (FLX) by photo- and biodegradation and shows similarities and differences in transformation products (TPs). TPs were identified using LC-high-resolution mass spectrometry with positive and negative electrospray ionization. In a sunlight
D C Thompson et al.
Chemico-biological interactions, 126(1), 1-14 (2000-05-29)
4-Trifluoromethylphenol (4-TFMP) was cytotoxic to precision-cut rat liver slices as indicated by loss of intracellular potassium. Intracellular glutathione levels decreased and fluoride ion levels increased in a time and concentration-dependent manner. The cytotoxicity of 4-TFMP did not appear to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service