Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3OC6H4OH
CAS Number:
Molecular Weight:
124.14
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
205-769-8
Beilstein/REAXYS Number:
507924
MDL number:
Assay:
99%
vapor density
4.3 (vs air)
vapor pressure
<0.01 mmHg ( 20 °C)
product line
ReagentPlus®
assay
99%
autoignition temp.
789 °F
bp
243 °C (lit.)
mp
55-57 °C (lit.)
SMILES string
COc1ccc(O)cc1
InChI
1S/C7H8O2/c1-9-7-4-2-6(8)3-5-7/h2-5,8H,1H3
InChI key
NWVVVBRKAWDGAB-UHFFFAOYSA-N
Gene Information
human ... TYR(7299)
Application
4-methoxyphenol may be used in the synthesis of butylated hydroxy anisoles via alkylation with methyl tert-butyl ether over a non-zeolitic solid acidic catalyst. This process is eco-friendly when compared to the Friedel-Crafts alkylation reaction. 4-MP may also react with aqueous nitrous acid to form 2-nitro-4-methoxyphenol and benzoquinone in varying ratios depending on the reaction conditions. 4-MP can be used as a building block in designing β-cyclodextrin 4-methoxyphenol conjugates that can be potential ligands for drug complexation.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under 4-Methoxyphenol
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chin-Wen Chen et al.
Polymers, 12(5) (2020-05-23)
Unsaturated poly (butylene adipate-co-butylene itaconate) (PBABI) copolyesters were synthesized through melt polymerization composed of 1,4-butanediol (BDO), adipic acid (AA), itaconic acid (IA) and 1,2,4,5-benzenetetracarboxylic acid (BTCA) as a cross-linking modifier. The melting point, crystallization and glass transition temperature of the
Nitration and oxidation of 4-methoxyphenol by nitrous acid in aqueous acid solution.
Beake B D
J. Chem. Soc. Perkin Trans. II, (2), 335-340 (1994)
Intermolecular interactions between doxorubicin and ?-cyclodextrin 4-methoxyphenol conjugates
Swiech O
The Journal of Physical Chemistry B, 116(6), 1765-1771 (2012)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| M18655-500G | 04061834041191 |
| M18655-100G | 04061835559510 |
| M18655-1KG | 04061834041184 |
| M18655-50LB | 04061832885964 |
| M18655-5G | 04061834041207 |
