Skip to Content
Merck
All Photos(1)

Key Documents

F13207

Sigma-Aldrich

4-Fluorophenol

99%

Synonym(s):

4-Hydroxyphenyl fluoride, p-Fluorophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4OH
CAS Number:
Molecular Weight:
112.10
Beilstein:
1362752
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

185 °C (lit.)

mp

43-46 °C (lit.)

SMILES string

Oc1ccc(F)cc1

InChI

1S/C6H5FO/c7-5-1-3-6(8)4-2-5/h1-4,8H

InChI key

RHMPLDJJXGPMEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Osman et al.
Chemico-biological interactions, 104(2-3), 147-164 (1997-05-02)
The present study shows that MP8 in the presence of H2O2 is able to catalyze the rupture of the stable carbon-fluorine bond of 4-fluorophenol, used as a model substrate for the oxidative dehalogenation reaction. 1,4-Benzoquinone was shown to be the
Alexandria Harkey et al.
Biotechnology letters, 34(9), 1725-1731 (2012-05-29)
Cytochrome P450(BM3)-F87G catalyzed the oxidative defluorination of 4-fluorophenol, followed by reduction of the resulting benzoquinone to hydroquinone via the NADPH P450-reductase activity of the enzyme. The k (cat) and K (m) for this reaction were 71 ± 5 min(-1) and
Manfred Lehnig et al.
Organic & biomolecular chemistry, 4(4), 721-729 (2006-02-10)
Peroxynitric acid (O2NOOH) nitrates L-tyrosine and related compounds at pH 2-5. During reaction with O2(15)NOOH in the probe of a 15N NMR spectrometer, the NMR signals of the nitration products of L-tyrosine, N-acetyl-L-tyrosine, 4-fluorophenol and 4-methoxyphenylacetic acid appear in emission
Lydie Coulombel et al.
Applied microbiology and biotechnology, 89(6), 1867-1875 (2010-11-09)
Escherichia coli cells, expressing 4-hydroxyphenylacetate 3-hydroxylase, fully transformed 4-halogenated phenols to their equivalent catechols as single products in shaken flasks. 4-Fluorophenol was transformed at a rate 1.6, 1.8, and 3.4-fold higher than the biotransformation of 4-chloro-, 4-bromo-, and 4-iodo-phenol, respectively.
Maria Isabel M Ferreira et al.
Applied microbiology and biotechnology, 78(4), 709-717 (2008-01-30)
A Gram-positive bacterial strain capable of aerobic biodegradation of 4-fluorophenol (4-FP) as the sole source of carbon and energy was isolated by selective enrichment from soil samples collected near an industrial site. The organism, designated strain IF1, was identified as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service