Skip to Content
Merck
All Photos(1)

Documents

164968

Sigma-Aldrich

5-Amino-4-imidazolecarboxamide hydrochloride

98%

Synonym(s):

4-Amino-5-imidazolecarboxamide hydrochloride, AICA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O · HCl
CAS Number:
Molecular Weight:
162.58
Beilstein:
3701645
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

250-252 °C (dec.) (lit.)

solubility

water: soluble 50 mg/mL, clear, light yellow

SMILES string

Cl[H].NC(=O)c1[nH]cnc1N

InChI

1S/C4H6N4O.ClH/c5-3-2(4(6)9)7-1-8-3;/h1H,5H2,(H2,6,9)(H,7,8);1H

InChI key

MXCUYSMIELHIQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Corrosion inhibition and adsorption characteristics of 5-amino-4-imidazolecarboxamide hydrochloride on aluminum in 1M HCl has been investigated.

Application

5-Amino-4-imidazolecarboxamide hydrochloride was used in the synthesis of heterocyclic compounds such as guanine, purines and pyrimidines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 56, 2139-2139 (1991)
Heterocycles, 34, 1133-1133 (1992)
Journal of Heterocyclic Chemistry, 30, 593-593 (1993)
Electrochemical and molecular dynamics simulation studies on the corrosion inhibition of aluminum in molar hydrochloric acid using some imidazole derivatives.
Khaled KF and Amin MA.
J. Appl. Electrochem., 39(12), 2553-2568 (2009)
Brandon Tran et al.
Cell chemical biology, 27(10), 1241-1249 (2020-08-17)
Riboswitches are mRNA domains that make gene-regulatory decisions upon binding their cognate ligands. Bacterial riboswitches that specifically recognize 5-aminoimidazole-4-carboxamide riboside 5'-monophosphate (ZMP) and 5'-triphosphate (ZTP) regulate genes involved in folate and purine metabolism. Now, we have developed synthetic ligands targeting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service