Skip to Content
Merck
All Photos(1)

Key Documents

T2551

Sigma-Aldrich

Monoclonal Anti-Tenascin antibody produced in mouse

clone BC-24, ascites fluid

Synonym(s):

Anti-Tenascin-N

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

BC-24, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:4,000 using protease-digested sections of adult human tissue.
immunohistochemistry (frozen sections): suitable
indirect ELISA: suitable
microarray: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNN(63923)

General description

Monoclonal Anti-Human Tenascin (mouse IgG1 isotype) is derived from the BC-24 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
Tenascin (TN) is a high molecular weight, multifunctional, extracellular matrix glycoprotein, expressed in association with mesenchymal-epithelial interactions during development and in the neovasculature and stroma of undifferentiated tumors. Human TN has 3 subunits of 190, 200 and 220 kD. TN has been independently discovered in a variety of species and tissue types, often in the basement membrane or intercellular spaces. The expression of TN is associated with development and growth, both normal and pathological, with restricted distribution in normal adult tissue. It is synthesized by fibroblasts, chondroblasts, osteocytes, smooth muscle cells and glial cells. Human and chicken TN contain an RGD sequence which may function in cell adhesion and it seems likely that TN mediates cell attachment through an RGD-dependent integrin receptor. Mouse TN does not contain an RGD sequence in the third type III repeat implicated in cell attachment, or in any other positions.

Specificity

The antibody recognizes an epitope within the N-terminal EGF-like (EGF-L) sequence of the molecule. It recognizes all human tenascin isoforms.

Immunogen

human tenascin.

Application

Monoclonal Anti-Tenascin antibody produced in mouse has been used for immunohistochemistry.
Mouse monoclonal clone BC-24 anti-Tenascin antibody is an essential tool for the localization, identification and studies on the role of tenascin in epithelial-mesenchymal and neuronal-glial interactions.

Biochem/physiol Actions

Tenascin (TN) is up-regulated by epithelial transforming growth factor β (TGF-β) in mesenchyme. TN in the stroma of cancerous tissue contributes to tumorigenesis and progression of metastasis. High molecular weight TN isoform, a stromal marker of breast cancer, over-expresses implicating cell proliferation and invasion. TN has a potential role in tissue restoration. Intracytoplasmic TN immunoreactivity has been detected in malignant melanomas and in lung carcinomas. TN expression, when reacted with monoclonal antibody, determines its localization in tumor cells and its association with epithelial-mesenchymal and neuronal-glial cell interactions.

Physical form

This product is supplied as ascites fluid containing 15 mM sodium azide

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G I Mason et al.
Annals of the rheumatic diseases, 59(3), 183-189 (2000-03-04)
To determine whether there was altered elaboration of non-collagenous extracellular matrix proteins or expression of TGFbeta isoforms and their receptors in salivary glands of patients with systemic sclerosis (SSc) and Raynaud's phenomenon (RP). Because of the possible role of mast
Changes in tenascin-C isoform expression in invasive and preinvasive breast disease
AdamS M, et al.
Cancer Research, 62(11), 3289-3297 (2002)
Tenascin-C in primary malignant melanoma of the skin
Ilmonen S, et al.
Histopathology, 45(4), 405-411 (2004)
Allegra G Hawkins et al.
Molecular & cellular proteomics : MCP, 17(5), 901-912 (2018-02-02)
Tumor: tumor microenvironment (TME) interactions are critical for tumor progression and the composition and structure of the local extracellular matrix (ECM) are key determinants of tumor metastasis. We recently reported that activation of Wnt/beta-catenin signaling in Ewing sarcoma cells induces
Expression of tenascin-C in various human brain tumors and its relevance for survival in patients with astrocytoma
Leins A, et al.
Cancer, 98(11), 2430-2439 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service