157341
3,5-Dichloroanisole
98%
Synonym(s):
1,3-Dichloro-5-methoxybenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Cl2C6H3OCH3
CAS Number:
Molecular Weight:
177.03
Beilstein:
1936395
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
mp
39-41 °C (dec.) (lit.)
functional group
chloro
SMILES string
COc1cc(Cl)cc(Cl)c1
InChI
1S/C7H6Cl2O/c1-10-7-3-5(8)2-6(9)4-7/h2-4H,1H3
InChI key
SSNXYMVLSOMJLU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
3,5-Dichloroanisole on nitration under mild conditions yields 3,5-dichloro-2,4,6-trinitroanisole. The 35Cl Nuclear Quadrupole Resonance (NQR) frequency and spin lattice relaxation time in 3,5-dichloroanisole has been measured.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
222.8 °F - closed cup
Flash Point(C)
106 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pressure Dependence of the Chlorine NQR in Three Solid Chloro Anisoles.
Suresh KS, et al.
Zeitschrift fur Naturforschung, A, 56(11), 764-772 (2001)
Preparation of 1, 3, 5-triamino-2, 4, 6-trinitrobenzene from 3, 5-dichloroanisole.
Ott DG and Benziger TM.
Journal of Energetic Materials, 5(3-4), 343-354 (1987)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service