Skip to Content
Merck
All Photos(3)

Key Documents

155837

Sigma-Aldrich

Tetrabutylammonium hydrogensulfate

97%

Synonym(s):

Tetrabutylammonium bisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
97%
form:
solid

Quality Level

Assay

97%

form

solid

mp

169-171 °C (lit.)

functional group

amine

SMILES string

OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tetrabutylammonium hydrogensulfate is an ammonium salt. It serves as solvent for the dissolution of ruthenium(II) complex fac-[Ru(CO)2(H2O)3(C(O)C2H5)][CF3SO3]. It participates as a cocatalyst in the reaction medium for the preparation of alternating polyketones and propionic acid.

Application

Tetrabutylammonium hydrogensulfate may be employed as a catalyst for the green synthesis of glycosyl 1,4-dihydropyridines.
Tetrabutylammonium hydrogensulfate may be employed as a catalyst for the green synthesis of glycosyl 1,4-dihydropyridines.
Tetrabutylammonium hydrogensulfate can serve as ion-pairing agent in the determination of the following using gas chromatography(GC) method :
  • linear alkylbenzenesulfonates(LASs)
  • acidic herbicides

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

348.1 °F - closed cup

Flash Point(C)

175.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Derivatization and solid-phase microextraction
Stashenko.EE and Martinez.RJ
TrAC, Trends in Analytical Chemistry, 23(8), 553-561 (2004)
Hydrocarboxylation and stepwise alternating oligomerization of carbon monoxide and ethylene catalyzed by cis-[Rh(CO)2(amine)2](PF6) complexes in a wet liquid tetrabutylammonium hydrogensulfate.
Pardey AJ, et al.
Polyhedron, 23(10), 1677-1683 (2004)
Tetrabutylammonium hydrogen sulfate catalyzed eco-friendly and efficient synthesis of glycosyl 1, 4-dihydropyridines.
Tewari N, et al.
Tetrahedron Letters, 45(49), 9011-9014 (2004)
New media in homogeneous catalysis: wet sodium or tetrabutylammonium hydrogensulfate salts for reppe syntheses catalyzed by a ru (ii) carbonyl complex.
Pardey AJ, et al.
Journal of Coordination Chemistry, 57(10), 871-882 (2004)
Environmental and bioanalytical applications of hollow fiber membrane liquid-phase microextraction: a review
Lee J, et al.
Analytica Chimica Acta, 624(2), 253-268 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service