Skip to Content
Merck
All Photos(1)

Documents

15661

Sigma-Aldrich

Boc-6-Ahx-OSu

≥98.0% (TLC)

Synonym(s):

6-(Boc-amino)caproic acid N-succinimidyl ester, 6-(Boc-amino)hexanoic acid N-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O6
CAS Number:
Molecular Weight:
328.36
Beilstein:
1505656
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

87-92 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCCCCC(=O)ON1C(=O)CCC1=O

InChI

1S/C15H24N2O6/c1-15(2,3)22-14(21)16-10-6-4-5-7-13(20)23-17-11(18)8-9-12(17)19/h4-10H2,1-3H3,(H,16,21)

InChI key

TYJPSIQEEXOQLC-UHFFFAOYSA-N

Other Notes

Linker for preparing fluoroscent probes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Juliano et al.
The journal of peptide research : official journal of the American Peptide Society, 53(2), 109-119 (1999-04-09)
We synthesized short chromogenic peptidyl-Arg-p-nitroanilides containing either (Galbeta)Ser or (Glcalpha,beta)Tyr at P2 or P3 sites as well as O-acetylated sugar moieties and studied their hydrolysis by bovine trypsin, papain, human tissue kallikrein and rat tonin. For comparison, the susceptibility to
M Adamczyk et al.
Steroids, 62(6), 462-467 (1997-06-01)
6-Oxoestradiol (2) was protected as its bis[(2-trimethylsilylethoxy)methyl] ether (4) and converted to the corresponding oxime (4). The oxime (4) on reduction with zinc in ethanol afforded the bis-SEM ether of 6-alpha-aminoestradiol (5) in 96% epimeric excess. Subsequently, 5 was hydrolyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service