Skip to Content
Merck
All Photos(1)

Documents

235385

Sigma-Aldrich

tert-Butyl nitrite

90%

Synonym(s):

1,1-Dimethylethyl nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CONO
CAS Number:
Molecular Weight:
103.12
Beilstein:
1209339
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.368 (lit.)

bp

61-63 °C (lit.)

solubility

alcohol: very soluble(lit.)
carbon disulfide: very soluble(lit.)
chloroform: very soluble(lit.)
diethyl ether: very soluble(lit.)
glycerol: insoluble (practically)(lit.)
water: slightly soluble(lit.)

density

0.867 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)ON=O

InChI

1S/C4H9NO2/c1-4(2,3)7-5-6/h1-3H3

InChI key

IOGXOCVLYRDXLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl nitrite (TBN) is an efficient NO source. TBN participates in photocatalyzed conversion of aryl- and heteroarylamines to selenides. It also participates in radical multifunctionalization reactions of aliphatic alkenes.

Application

tert-Butyl nitrite has been used as reagent:
  • for diazotization and nitros­ation of alcohols, thiols, amines and cycloalkanes
  • for the preparation of aryl azides from aryl amines

Other Notes

remainder 2-methyl-2-propanol

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 42, 3494-3494 (1977)
tert-Butyl Nitrite.
Liu Y.
Synlett, 2011(08), 1184-1185 (2011)
Daisuke Hirose et al.
Beilstein journal of organic chemistry, 9, 1713-1717 (2013-09-26)
Water induces a change in the product of radical multifunctionalization reactions of aliphatic alkenes involving an sp(3) C-H functionalization by an 1,5-hydrogen shift using tert-butyl nitrite and molecular oxygen. The reaction without water, reported previously, gives nitrated γ-lactols, whereas the
The Journal of Organic Chemistry, 42, 2426-2426 (1977)
Debasish Kundu et al.
Organic letters, 16(6), 1814-1817 (2014-03-14)
A novel strategy for the direct conversion of aryl- and heteroarylamines to selenides has been developed via diazotization of amines with tert-butyl nitrite in neutral medium followed by reaction with diaryl/diheteroaryl/dialkyl diselenides in one pot under photocatalysis at room temperature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service