Skip to Content
Merck
All Photos(1)

Key Documents

151610

Sigma-Aldrich

DL-3,4-Dihydroxymandelic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH(OH)CO2H
CAS Number:
Molecular Weight:
184.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

136-137 °C (dec.) (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

OC(C(O)=O)c1ccc(O)c(O)c1

InChI

1S/C8H8O5/c9-5-2-1-4(3-6(5)10)7(11)8(12)13/h1-3,7,9-11H,(H,12,13)

InChI key

RGHMISIYKIHAJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Metabolite of norepinephrine.

Application

DL-3,4-Dihydroxymandelic acid was used in the simultaneous analysis of 4-hydroxy-3-methoxymandelic acid and 4-hydroxy- 3-methoxyphenylacetic acid in urine. It was also used to study the changes in body temperature.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Y Li et al.
Analytical biochemistry, 190(2), 354-359 (1990-11-01)
A quantitative assay for the diphenol oxidase activity of tyrosinase (EC 1.14.18.1) using high-pressure liquid chromatography with electrochemical detection is described. The assay is based on the observation (M. Sugumaran, 1986, Biochemistry 25, 4489-4492) that tyrosinase catalyzes the oxidative decarboxylation
M Kawamura et al.
Journal of the autonomic nervous system, 66(3), 145-148 (1997-12-24)
After norepinephrine (NE) is deaminated by monoamine oxidase (MAO), the aldehyde formed is either metabolized to 3,4-dihydroxy-mandelic acid (DHMA) by aldehyde dehydrogenase or is converted to 3,4-dihydroxyphenylglycol (DHPG) by aldehyde or aldose reductase. The present study examined the effects of
S J Soldin et al.
Clinical chemistry, 26(2), 291-294 (1980-02-01)
We describe a rapid, reliable "high-performance" liquid-chromatographic method of simultaneously analyzing for 4-hydroxy-3-methoxymandelic acid (I) and 4-hydroxy-3-methoxyphenylacetic acid (II) in urine. Paired-ion chromatography and amperometric detection are used in the method. A 5-mL aliquot of urine is adjusted to pH
M Sugumaran et al.
The Biochemical journal, 281 ( Pt 2), 353-357 (1992-01-15)
Mushroom tyrosinase, which is known to convert a variety of o-diphenols into o-benzoquinones, has been shown to catalyse an unusual oxidative decarboxylation of 3,4-dihydroxymandelic acid to 3,4-dihydroxybenzaldehyde [Sugumaran (1986) Biochemistry 25, 4489-4492]. The mechanism of this reaction was re-investigated. Although
J N Rodríguez-López et al.
Analytical biochemistry, 195(2), 369-374 (1991-06-01)
A continuous spectrophotometric method for the rapid determination of diphenolase activity of tyrosinase is described. It uses 3,4-dihydroxymandelic acid (DOMA) as the substrate of tyrosinase and measures the final product, 3,4-dihydroxybenzaldehyde (DOBA). The spectrum of this product shows a bathochromic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service