Skip to Content
Merck

B6295

Benzalkonium chloride

BioXtra

Synonym(s):

Alkylbenzyldimethylammonium chloride, Alkyldimethylbenzylammonium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH2N(CH3)2RCl (R=C8H17 to C18H37)
CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
264-151-6
Beilstein/REAXYS Number:
4062599
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

cationic

Quality Segment

product line

BioXtra

assay

95.0-105.0% (calculated on dry substance)

form

(Solid or Semi-Solid or Viscous Liquid)

impurities

≤0.001% Phosphate, ≤0.1% Insoluble matter, ≤10% water

ign. residue

≤2%

solubility

water: insoluble

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%, Ca: ≤0.0005%, Cu: ≤0.0005%, Fe: ≤0.0005%, K: ≤0.005%, Mg: ≤0.0005%, Na: ≤0.02%, Pb: ≤0.001%, Zn: ≤0.0005%

SMILES string

[Cl-].CCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C19H34N.ClH/c1-4-5-6-7-8-9-10-14-17-20(2,3)18-19-15-12-11-13-16-19;/h11-13,15-16H,4-10,14,17-18H2,1-3H3;1H/q+1;/p-1

InChI key

TTZLKXKJIMOHHG-UHFFFAOYSA-M

General description

Benzalkonium chloride is a cationic detergent that is a member of quaternary ammonium compounds (QACs).

Application

Benzalkonium chloride has been used in a study to assess its action on epithelia conjunctival cells in vitro. It has also been used in a study to investigate its effects on living rabbit cornea by in vivo Tandem scanning confocal microscopy.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.


Still not finding the right product?

Explore all of our products under Benzalkonium chloride


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library