112720
N,N-Diethylethylenediamine
≥99%
Synonym(s):
2-Diethylaminoethylamine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(C2H5)2NCH2CH2NH2
CAS Number:
Molecular Weight:
116.20
Beilstein:
605299
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99%
refractive index
n20/D 1.436 (lit.)
bp
145-147 °C (lit.)
density
0.827 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CCN(CC)CCN
InChI
1S/C6H16N2/c1-3-8(4-2)6-5-7/h3-7H2,1-2H3
InChI key
UDGSVBYJWHOHNN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
N,N-Diethylethylenediamine was used in the oxidative amidation of hydrogen phosphonate diesters in the synthesis of oligonucleotides.
N,N-diethylethylenediamine was used to develop melanin targeted PET and single-photon emission computed tomography (SPECT) imaging agents for melanoma.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Flam. Liq. 3 - Skin Corr. 1A
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
89.6 °F - closed cup
Flash Point(C)
32 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J M Dagle et al.
Nucleic acids research, 28(10), 2153-2157 (2000-04-25)
We have designed a new class of modified antisense oligodeoxyribonucleotides (ODN) consisting of a central contiguous stretch of 6-8 unmodified nucleotides flanked by 3'- and 5'-regions containing several nucleotides joined by cationic internucleoside linkages. The positive charge results from modification
Hongguang Liu et al.
Molecular pharmaceutics, 10(9), 3384-3391 (2013-08-10)
Radiofluorinated benzamide and nicotinamide analogues are promising molecular probes for the positron emission tomography (PET) imaging of melanoma. Compounds containing aromatic (benzene or pyridine) and N,N-diethylethylenediamine groups have been successfully used for development of melanin targeted PET and single-photon emission
Diana Salakhieva et al.
International journal of pharmaceutics, 517(1-2), 234-246 (2016-12-10)
A series of 14 cationic derivatives of poly(aspartic acid) i.e. cationic polyaspartamides with different (dialkylamino)alkyl and alkyl or hydroxyalkyl side groups was synthesized by nucleophilic addition on polysuccinimide. The resulting polyaspartamides have moderate amphiphilic properties. Relationships between the structure and
Vincenzo Taresco et al.
Materials science & engineering. C, Materials for biological applications, 52, 72-81 (2015-05-09)
The application of magnetic nanoparticles (MNPs) in medicine is considered much promising especially because they can be handled and directed to specific body sites by external magnetic fields. MNPs have been investigated in magnetic resonance imaging, hyperthermia and drug targeting.
Feng-Yun J Huang et al.
International journal of nanomedicine, 10, 463-473 (2015-01-28)
In this study, the (188)Re-labeled PEGylated nanoliposome ((188)Re-liposome) was prepared and evaluated as a therapeutic agent for glioma. The reporter cell line, F98(luc) was prepared via Lentivector expression kit system and used to set up the orthotopic glioma-bearing rat model
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service