127000
N-Ethylethylenediamine
98%
Synonym(s):
2-Ethylaminoethylamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C2H5NHCH2CH2NH2
CAS Number:
Molecular Weight:
88.15
Beilstein:
1098272
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.439 (lit.)
bp
128-130 °C (lit.)
density
0.837 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CCNCCN
InChI
1S/C4H12N2/c1-2-6-4-3-5/h6H,2-5H2,1H3
InChI key
SCZVXVGZMZRGRU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
N-Ethylethylenediamine undergoes condensation with 2-hydroxy-3-methoxybenzaldehyde. It also forms organometallic ruthenium(II) complexes that inhibit the growth in human ovarian cancer cell lines.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Resp. Sens. 1 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
50.0 °F - closed cup
Flash Point(C)
10 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
R E Morris et al.
Journal of medicinal chemistry, 44(22), 3616-3621 (2001-10-19)
Inhibition of the growth of the human ovarian cancer cell line A2780 by organometallic ruthenium(II) complexes of the type [(eta(6)-arene)Ru(X)(Y)(Z)], where arene is benzene or substituted benzene, X, Y, and Z are halide, acetonitrile, or isonicotinamide, or X,Y is ethylenediamine
L Narasimha Murthy et al.
Journal of food science and technology, 58(2), 427-436 (2021-02-12)
Present study evaluates the effect of calcium and heat setting on gel characteristics of lesser sardine (Sardinella fimbriata) surimi. Total seven formulations were made namely, S. fimbriata minced meat without heat setting (A1), minced meat with heat setting (A2), S.
Lihua Hu et al.
Scientific reports, 7(1), 3611-3611 (2017-06-18)
Water-soluble hyperbranched polyamine functionalized multiwalled carbon nanotubes nanocomposite (WHPA-OMCNT) was successfully prepared and applied to water remediation in this paper. WHPA-OMCNT was characterized by Fourier transform infrared spectroscopy (FTIR), thermogravimetric analysis (TGA), zeta potential, scanning electron microscopy (SEM) and transmission
Sandip Mukherjee et al.
Dalton transactions (Cambridge, England : 2003), 42(11), 4019-4030 (2013-01-24)
Three new copper-azido complexes [Cu(4)(N(3))(8)(L(1))(2)](n) (1), [Cu(4)(N(3))(6)(L(2))(2)(H(2)O)(2)] (2), and [Cu(4)(N(3))(6)(L(3))(2)](n) (3) [L(1) is the imine resulting from the condensation of pyridine-2-carboxaldehyde with N-methylethylenediamine, HL(2) and HL(3) are the condensation products of 2-hydroxy-3-methoxybenzaldehyde with N,N-diethylethylenediamine and N-ethylethylenediamine respectively] have been synthesized
Guo-Ping Cheng et al.
Acta chimica Slovenica, 64(1), 261-265 (2017-04-06)
A rarely seen phenolato and azido-bridged polymeric cadmium(II) complex derived from the Schiff base ligand 2-bromo-6-[(2-isopropylaminoethylimino)methyl]phenol (HL) has been prepared and characterized by elemental analysis, IR spectroscopy, and single crystal X-ray diffraction. The Schiff base ligand coordinates to the Cd
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service