Skip to Content
Merck
All Photos(1)

Key Documents

259268

Sigma-Aldrich

Hexaethylene glycol

97%, average MN 300

Synonym(s):

Polyethylene glycol, 3,6,9,12,15-Pentaoxaheptadecane-1,17-diol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)6OH
CAS Number:
Molecular Weight:
282.33
Beilstein:
1638281
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Hexaethylene glycol, 97%

Quality Level

Assay

97%

form

liquid

mol wt

average Mn 300

refractive index

n20/D 1.465 (lit.)

bp

217 °C/4 mmHg (lit.)

mp

5-7 °C (lit.)

density

1.127 g/mL at 25 °C (lit.)

Ω-end

hydroxyl

α-end

hydroxyl

SMILES string

OCCOCCOCCOCCOCCOCCO

InChI

1S/C12H26O7/c13-1-3-15-5-7-17-9-11-19-12-10-18-8-6-16-4-2-14/h13-14H,1-12H2

InChI key

IIRDTKBZINWQAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Substrate employed in the synthesis of binaphthol-based macrocyclic ethers using intramolecular oxidative coupling with CuCl(OH)-TMEDA.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 8563-8563 (2006)
Xin Li et al.
Analytical chemistry, 84(6), 2609-2613 (2012-03-03)
Lipophilicity of chemicals and drug candidates is normally described in terms of octanol/water partitioning and log P. We investigated an alternate approach to lipophilicity determination using a mimic of an alkyl alcohol with compound partitioning quantified using acoustic sensing. A
M Durand et al.
Biochemistry, 35(28), 9133-9139 (1996-07-16)
The interaction of the minor groove binding drug distamycin with the T-A-T triple helix and the A-T double helix was studied using circular dichroism spectroscopy and thermal denaturation. The triple helix was made by the oligonucleotide (dA)12-x-(dT)12-x-(dT)12, where x is
O Bornet et al.
Journal of biomolecular NMR, 4(4), 575-580 (1994-07-01)
We present NMR studies of an intramolecular triple helix, the three strands of which have been linked by a hexaethylene glycol chain. To overcome the generally encountered difficulties of assignment in the homo-pyrimidine strands, the carbon Cl' of the pyrimidines
Ewelina Burakowska et al.
Small (Weinheim an der Bergstrasse, Germany), 5(19), 2199-2204 (2009-07-03)
A convenient methodology for the synthesis of photolabile crosslinked hyperbranched polyglycerol nanocapsules is presented. These nanocarriers selectively and efficiently bind ionic guest molecules. The stability of the host-guest complexes formed depends on the counterion of the guest molecules. Moreover, the

Articles

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service