Merck
  • Effects of thymoquinone on testicular structure and sperm production in male obese rats.
Effects of thymoquinone on testicular structure and sperm production in male obese rats.
Systems biology in reproductive medicine (2015-06-05)
Nur Hande Tüfek, Muhammad Eyüp Altunkaynak, Berrin Zuhal Altunkaynak, Süleyman Kaplan
ABSTRACT
Thymoquinone (TQ) is a phytochemical compound found in the plant Nigella sativa. It has antioxidant and anti-cancer effects. This study investigated the effects of TQ on obesity and testicular structure of high-fat-diet (HFD) fed rats. Obese control (OC) and obese thymoquinone (OT) groups were fed a special diet containing 40% of total calories from fat. Non-obese control (NC) and non-thymoquinone (NT) groups were fed a standard diet for nine weeks. Then, intraperitoneal TQ injections were carried out to the OT and NT groups for six weeks and testes were removed. Catalase and myeloperoxidase activity were determined in rat testis tissue. Stereological, histopathological, and immunohistochemical changes were evaluated in the testes of the rats. In stereological studies, mean volumes of testis and seminiferous tubules, the number of spermatogenic cells and also Leydig cells in the OC group were reduced, but these values significantly increased in the OT group. Apoptotic cells were observed in the OC group in comparison to the OT group. The number of healthy sperms were reduced in the OC group, whereas the majority showed anomalies in the head, neck, and tail. The number of healthy sperm was increased and the anomalies significantly reduced by using TQ in both the NT, and especially the OT group. TQ like antioxidants may improve fertility by means of increasing the healthy sperm number and preventing sperm anomalies.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Anti-Luteinizing Hormone/Choriogonadotropin Receptor Antibody, Chemicon®, from rabbit