Merck
  • Development and validation of a dynamic range-extended LC-MS/MS multi-analyte method for 11 different postmortem matrices for redistribution studies applying solvent calibration and additional (13)C isotope monitoring.
Development and validation of a dynamic range-extended LC-MS/MS multi-analyte method for 11 different postmortem matrices for redistribution studies applying solvent calibration and additional (13)C isotope monitoring.
Analytical and bioanalytical chemistry (2015-09-24)
Sandra N Staeheli, Michael Poetzsch, Thomas Kraemer, Andrea E Steuer
ABSTRACT
Postmortem redistribution (PMR) is one of numerous problems in postmortem toxicology making correct interpretation of measured drug concentrations difficult or even impossible. Time-dependent PMR in peripheral blood and especially in tissue samples is still under-explored. For further investigation, an easy applicable method for the simultaneous quantitation of over 80 forensically relevant compounds in 11 different postmortem matrices should be developed and validated overcoming the challenges of high inter-matrix and intra-matrix concentration variances. Biopsy samples (20 mg) or body fluids (20 μL) were spiked with an analyte mix and deuterated internal standards, extracted by liquid-liquid extraction, and analyzed by liquid chromatography tandem mass spectrometry (LC-MS/MS). For highest applicability, an easy solvent calibration was used. Furthermore, time-consuming dilution of high concentration samples showing detector saturation was circumvented by two overlapping calibration curves using (12)C isotope monitoring for low concentrations and (13)C isotopes for high concentration, respectively. The method was validated according to international guidelines with modifications. Matrix effects and extraction efficiency were strongly matrix and analyte dependent. In general, brain and adipose tissue produced the highest matrix effects, whereas cerebrospinal fluid showed the least matrix effects. Accuracy and precision results were rather matrix independent with some exceptions. Despite using an external solvent calibration, the accuracy requirements were fulfilled for 66 to 81 % of the 83 analytes. Depending on the matrix, 75-93 % of the analytes showed intra-day precisions at <20 %. (12)C and (13)C calibrations gave comparable results and proved to be a useful tool in expanding the dynamic range.
Product Description
Supelco
Clonazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetal, ≥98%, FG
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Zolpidem, ≥98% (HPLC), solid
Sigma-Aldrich
Clonazepam, powder
Sigma-Aldrich
Zopiclone
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetal, natural, ≥97%, FG
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)