Skip to Content
Merck
All Photos(1)

Documents

C-907

Supelco

Clonazepam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10ClN3O3
CAS Number:
Molecular Weight:
315.71
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1ccc2NC(=O)CN=C(c3ccccc3Cl)c2c1

InChI

1S/C15H10ClN3O3/c16-12-4-2-1-3-10(12)15-11-7-9(19(21)22)5-6-13(11)18-14(20)8-17-15/h1-7H,8H2,(H,18,20)

InChI key

DGBIGWXXNGSACT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clonazepam, marketed under trade names such as Klonopin and Rivotril, is a potent, long-acting benzodiazepine with anxiolytic, anticonvulsant, muscle relaxant and hypnotic properties. Clonazepam is prescribed for the treatment of epilepsy. This certified Snap-N-Spike® solution is suitable for use in GC/MS or LC/MS applications in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.

Application


  • Clonazepam research solution: Research on chemometric methods for determining the dissociation constants of benzodiazepine derivatives, including clonazepam, showcases its critical role in improving the precision of pharmaceutical analyses. This method enhances the understanding of clonazepam′s interactions in various conditions, aiding in its application in scientific research and drug formulation (Shayesteh et al., 2015).

  • High-purity clonazepam liquid for scientific study: A study employing solubility prediction models for drugs, including clonazepam, in mixed solvents provides significant insights into its solubility behavior. This knowledge is crucial for optimizing drug formulations and enhancing bioavailability in pharmaceutical preparations (Jouyban et al., 2011).

  • Clonazepam as a GABA receptor agonist in lab experiments: An investigation into multibenzodiazepine determination in human plasma emphasizes the importance of clonazepam in clinical and pharmaceutical research, particularly its role in affecting GABA receptors, crucial for understanding its pharmacological impacts (Borges et al., 2009).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taeko Sasai et al.
Parkinsonism & related disorders, 19(2), 153-157 (2012-09-20)
The clinical effectiveness of pramipexole (PPX), a candidate treatment for idiopathic rapid eye movement sleep behavior disorder (iRBD), varies among individuals. This study investigates factors associated with PPX effectiveness for treating RBD symptoms. Ninety-eight consecutive patients with RBD who had
John C Oakley et al.
The Journal of pharmacology and experimental therapeutics, 345(2), 215-224 (2013-02-21)
Seizures remain uncontrolled in 30% of patients with epilepsy, even with concurrent use of multiple drugs, and uncontrolled seizures result in increased morbidity and mortality. An extreme example is Dravet syndrome (DS), an infantile-onset severe epilepsy caused by heterozygous loss
Shigeru Morishita
Human psychopharmacology, 24(3), 191-198 (2009-03-31)
Clonazepam, first used for seizure disorders, is now increasingly used to treat affective disorders. We summarize the use of clonazepam to improve the management of depression. Clonazepam is useful for treatment-resistant and/or protracted depression, as well as for acceleration of
Stuart J McCarter et al.
Sleep medicine, 14(3), 237-242 (2013-01-29)
REM sleep behavior disorder (RBD) is usually characterized by potentially injurious dream enactment behaviors (DEB). RBD treatment aims to reduce DEBs and prevent injury, but outcomes require further elucidation. We surveyed RBD patients to describe longitudinal treatment outcomes with melatonin
Raffaele Ferri et al.
Sleep medicine, 14(1), 24-29 (2012-10-27)
To analyze the differences in sleep structure and nocturnal motor activity between drug-free REM sleep behavior disorder (RBD) patients and those under therapy with clonazepam, and to evaluate the long-term longitudinal changes under continued therapy with clonazepam. Fifty-seven consecutive iRBD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service