Skip to Content
Merck
All Photos(1)

Key Documents

W200204

Sigma-Aldrich

Acetal

≥98%, FG

Synonym(s):

Acetaldehyde diethyl acetal, 1,1-Diethoxyethane, Acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OC2H5)2
CAS Number:
Molecular Weight:
118.17
FEMA Number:
2002
Beilstein:
1098310
EC Number:
Council of Europe no.:
35
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.001
NACRES:
NA.21
Organoleptic:
green; ethereal; vegetable; earthy; sweet
grade:
FG
Fragrance grade
Halal
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

4.1 (vs air)

vapor pressure

20 mmHg ( 20 °C)
8 mmHg ( 8 °C)

Assay

≥98%

autoignition temp.

446 °F

expl. lim.

10.4 %

refractive index

n20/D 1.379-1.383 (lit.)

bp

102 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; ethereal; vegetable; earthy; sweet

SMILES string

CCOC(C)OCC

InChI

1S/C6H14O2/c1-4-7-6(3)8-5-2/h6H,4-5H2,1-3H3

InChI key

DHKHKXVYLBGOIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Acetal is a volatile flavor compound mainly found in grapefruit essence, Durio zibethinus fruit pulp and grape pomace distillate.

Application


  • Synthesis of Diverse Allyl Sulfone Derivatives via Sequential Hydroalkoxylation of 1,3-Enynes.: This paper details a method for synthesizing allyl sulfone derivatives, which are valuable in various chemical manufacturing processes, through the controlled reaction of acetals (Kumar and Shah, 2024).


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<68.0 °F - closed cup

Flash Point(C)

< 20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile constituents and fatty acid composition of lipids in Durio zibethinus.
Moser R, et al.
Phytochemistry, 19(1), 79-81 (1980)
Analysis of volatile flavor constituents from grapefruit essence.
Moshonas MG & Shaw PE
Journal of Agricultural and Food Chemistry, 19(1), 119-120 (1971)
Fast determination of principal volatile compounds in distilled spirits.
Lopez-Vazquez C, et al.
Food Control, 21(11), 1436-1441 (2010)
Geelsu Hwang et al.
Biofouling, 30(9), 1079-1091 (2014-10-31)
Well-established biofilms formed by Streptococcus mutans via exopolysaccharide matrix synthesis are firmly attached to tooth surfaces. Enhanced understanding of the physical properties of mature biofilms may lead to improved approaches to detaching or disassembling these highly organized and adhesive structures.
A J Cappadoro et al.
Animal reproduction science, 158, 104-108 (2015-05-30)
The purpose of this study was to develop a new injection-molded intravaginal insert manufactured from ethylene-vinyl acetate containing progesterone for a 7-day insertion period in cattle. The manufacturing process resulted in a reduction in the residual drug compared to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service