Merck
  • Achiral-chiral two-dimensional chromatography of free amino acids in milk: A promising tool for detecting different levels of mastitis in cows.
Achiral-chiral two-dimensional chromatography of free amino acids in milk: A promising tool for detecting different levels of mastitis in cows.
Journal of pharmaceutical and biomedical analysis (2015-01-27)
Federica Ianni, Roccaldo Sardella, Antonella Lisanti, Antimo Gioiello, Beniamino Terzo Cenci Goga, Wolfgang Lindner, Benedetto Natalini
ABSTRACT
In two-dimensional HPLC (2D-HPLC) "heart-cut" applications, two columns are connected in series via a switching valve and volume fractions from the "primary" column are re-injected on the "secondary" column. The heart-cut 2D-HPLC system here described was implemented by connecting a reversed-phase (RP) column (first dimension) to a chiral column (second dimension) containing a quinidine-based chiral stationary phase. The system was used to evaluate the change in the enantiomeric excess value of dansylated (Dns) amino acids (AAs) in milk samples from two cows with different "California Mastitis Test" scores: negative test for sample 1, positive for sample 2. Apart from the co-elution of Dns-Arg/Dns-Gly and the reduced chemoselectivity for Dns-Leu/Dns-allo-Ile, the optimized achiral RP method distinguished the remaining standard Dns-AAs. Dns-AAs were identified in the chromatograms of the real samples, and in higher concentration Dns-Ala, Dns-Arg, Dns-Asp, Dns-Glu, Dns-Ile, Dns-Leu, Dns-Phe and Dns-Val. Except Dns-Arg, the chiral column enabled the RP enantioseparation of all the other compounds (α and RS values up to 1.65 and 8.63, respectively, for Dns-Phe). In sample 2, the amounts of Dns-d-AAs were rather elevated, in particular for Dns-Ala and Dns-Asp. Instead, for sample 1, D-isomers were detected for Dns-Ala, Dns-Glu and Dns-Leu. The proposed 2D-HPLC method could be useful for the identification of clinical mastitis difficult to be diagnosed. Moreover, the eventual progressive reduction of D-AAs levels with the degree of sub-clinical mastitis could allow the building of mathematical models to use for the diagnosis of early stages of mastitis.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%