Merck
  • Sustained reduction in methane production from long-term addition of 3-nitrooxypropanol to a beef cattle diet.
Sustained reduction in methane production from long-term addition of 3-nitrooxypropanol to a beef cattle diet.
Journal of animal science (2015-05-29)
A Romero-Perez, E K Okine, S M McGinn, L L Guan, M Oba, S M Duval, M Kindermann, K A Beauchemin
ABSTRACT
The objective was to evaluate whether long-term addition of 3-nitrooxypropanol (NOP) to a beef cattle diet results in a sustained reduction in enteric CH4 emissions in beef cattle. Eight ruminally cannulated heifers (637 ± 16.2 kg BW) were used in a completely randomized design with 2 treatments: Control (0 g/d of NOP) and NOP (2 g/d of NOP). Treatments were mixed by hand into the total mixed ration (60% forage, DM basis) at feeding time. Feed offered was restricted to 65% of ad libitum DMI (slightly over maintenance energy intake) and provided once per day. The duration of the experiment was 146 d, including an initial 18-d covariate period without NOP use; a 112-d treatment period with NOP addition to the diet, divided into four 28-d time intervals (d 1 to 28, 29 to 56, 57 to 84, and 85 to 112); and a final 16-d recovery period without NOP use. During the covariate period and at the end of each interval and the end of the recovery period, CH4 was measured for 3 d using whole animal metabolic chambers. The concentration of VFA was measured in rumen fluid samples collected 0, 3, and 6 h after feeding, and the microbial population was evaluated using rumen samples collected 3 h after feeding on d 12 of the covariate period, d 22 of each interval within the treatment period, and d 8 of the recovery period. Average DMI for the experiment was 7.04 ± 0.27 kg. Methane emissions were reduced by 59.2% when NOP was used (9.16 vs. 22.46 g/kg DMI; P < 0.01). Total VFA concentrations were not affected (P = 0.12); however, molar proportion of acetate was reduced and that for propionate increased when NOP was added (P < 0.01), which reduced the acetate to propionate ratio (3.0 vs. 4.0; P < 0.01). The total copy number of the 16S rRNA gene of total bacteria was not affected (P = 0.50) by NOP, but the copy number of the 16S rRNA gene of methanogens was reduced (P < 0.01) and the copy number of the 18S rRNA gene of protozoa was increased (P = 0.03). The residual effect of NOP for most of the variables studied was not observed or was minimal during the recovery period. These results demonstrated that the addition of NOP to a diet for beef cattle caused a sustained decrease of methanogenesis, with no sign of adaptation, and that these effects were reversed once NOP addition was discontinued
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, foil, 8x8mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon