Skip to Content
Merck
All Photos(2)

Key Documents

W1503

Sigma-Aldrich

Water

sterile filtered, suitable for embryo cell culture, BioXtra

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75
technique(s):
cell culture | embryo: suitable
bp:
100 °C (lit.)
vapor pressure:
3 mmHg
Pricing and availability is not currently available.

Product Name

Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture

vapor density

<1 (vs air)

Quality Level

vapor pressure

3 mmHg

sterility

sterile-filtered

product line

BioXtra

form

liquid

technique(s)

cell culture | embryo: suitable

impurities

endotoxin, tested

refractive index

n20/D 1.34 (lit.)

pH

5-7

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Water has been used in the preparation of mouse sperm cryopreservation medium, for suspension of particulate samples in various studies
Water is suitable for use in embryo manipulation.
Water has been used:
  • in toxicological analyses for the suspension of emission particles[1][2]
  • for suspension of particulate samples in various studies[3]
  • in cell cultivation and exposure[4]
  • for washing cells in calcium free water during the detection of intracellular calcium changes[5]

Other Notes

Easily compare specifications for water products with the water specification table.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro immunotoxic and genotoxic activities of particles emitted from two different small-scale wood combustion appliances.
Maija Tapanainen et al.
Atmospheric Environment, 45, 7546-7554 (2011)
Tiina Torvela et al.
Chemical research in toxicology, 27(9), 1516-1527 (2014-07-27)
Multiple studies show that particulate mass (PM) generated from incomplete wood combustion may induce adverse health issues in humans. Previous findings have shown that also the PM from efficient wood combustion may induce enhanced production of reactive oxygen species (ROS)
In vitro toxicological characterization of particulate emissions from residential biomass heating systems based on old and new technologies.
Pasi I. Jalava et al.
Atmospheric Environment, 50, 24-35 (2012)
G Morgan et al.
Cell death and differentiation, 6(1), 36-47 (1999-04-14)
A mutant T-cell hybridoma line named mutant 51 was developed that, unlike the parental line, did not die after T-cell receptor (TCR) engagement and demonstrated reduced death in response to dexamethasone. Intracellular calcium measurements showed that available calcium stores were
Jarno Ruusunen et al.
Analytical and bioanalytical chemistry, 401(10), 3183-3195 (2011-10-01)
Several studies have shown that combustion-derived fine particles cause adverse health effects. Previous toxicological studies on combustion-derived fine particles have rarely involved multiple endpoints and a detailed characterization of chemical composition. In this study, we developed a novel particle sampling

Articles

Mouse embryo media and embryo validated reagents for transgenic mouse embryo culture

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service