Skip to Content
Merck
All Photos(5)

Documents

699632

Sigma-Aldrich

Carbon, mesoporous

greener alternative

nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined

Synonym(s):

Graphitized carbon black

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

mol wt

Mw 12.011 g/mol

composition

C

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

150-250 m2/g

pore size

0.342 cm3/g total pore volume (typical)
64 Å average pore diameter (typical)

bp

4827 °C

mp

3654-3697 °C

density

0.056 g/cm3 (Packing)
1.887 g/cm3 (absolute, typical)

application(s)

battery manufacturing

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Carbon, mesoporous can be used for a variety of applications such as fuel cell, lithium ion batteries, lithium sulfur batteries, double-layer capacitor and hydrogen evolution.
It was used for a comparative study of the toxicity effect of carbon nanoparticles on C57BL/6 mice.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of rattle-type magnetic mesoporous carbon spheres and their highly efficient adsorption and separation
Yin Y, et al.
Journal of Colloid and Interface Science, 361(2), 527-533 (2011)
Metallic WO2-carbon mesoporous nanowires as highly efficient electrocatalysts for hydrogen evolution reaction
Wu R, et al.
Journal of the American Chemical Society, 137(22), 6983-6986 (2015)
Core-shell structured silicon nanoparticles@ TiO2-x/carbon mesoporous microfiber composite as a safe and high-performance lithium-ion battery anode
Jeong G, et al.
ACS Nano, 8(3), 2977-2985 (2014)
Traditional vs. Non-Traditional Materials for Carbon Paste Electrode
Preparation: On Relation between the Composition and Properties,
Mikysek T, et al.
Sensing in Electroanalysis, 8, 133-141 null
Preparation of mesoporous carbon by freeze drying
Tamon H, et al.
Carbon, 37(12), 2049-2055 (1999)

Articles

Mesoporous Materials include a range of high surface area porous silicates with applications in gas adsorption, drug delivery, diagnostics and catalysis.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service