Skip to Content
Merck
All Photos(4)

Key Documents

702110

Sigma-Aldrich

Carbon, mesoporous

greener alternative

hydrophilic pore surface

Synonym(s):

Starbon 300

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

greener alternative product characteristics

Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

>48.8 m2/g , BET

pore size

>0.13 cm3/g mesoporosity

bp

4827 °C

mp

3654-3697 °C

application(s)

battery manufacturing

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Free-flowing micron-sized powder
Functionality by FITR: OH, C=O, and C=C
Starbon is mesoporous carbon synthesized by the carbonization of expanded mesoporous starch at high temperatures. Amylose and amylopectin polymer chains in starch self assemble to form large mesoporous structures. Starbons exhibit tunable surface energies and high volume. It can be readily functionalized. Mesoporous carbon may have a hydrophilic pore surface and a BET surface area of 300 m2/g. The mean particle size of this graphitized carbon is 5 μm.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Carbon, mesoporous can be used in the esterification of oleic acid and lauric acid for the formation of biofuels. It forms a composite with nanoscale zero valent iron particles for the removal of arsenic from groundwater.
It was used for a comparative study of the toxicity effects of various carbon nanoparticles on C57BL/6 mice.

Legal Information

Starbon is a trademark of University of York

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Starbons?: Cooking up Nanostructured Mesoporous Materials
Budarin V, et al.
Material Matters, 4(1), 19-19 (2009)
Esterification of lauric acid with methanol using sulfonated Starbons
Mena-Duran Cinthia J and Macquarrie DJ
research journal of chemistry and environment, 18, 1-6 (2014)
Functionalized single-walled carbon nanotubes cause reversible
acute lung injury and induce fibrosis in mice
Xhang Y, et al.
Journal of Molecular Medicine, 91, 117-128 (2013)
Review of toxicity studies of carbon nanotubes
Kobayashi N, et al.
Journal of Occupational Health, 59(5), 394-407 (2017)
Water-tolerant Ru-Starbon
Luque R and Clark JH
Catalysis Communications, 11(10), 928-931 (2010)

Articles

Our strategy is to synthesize mesoporous carbonaceous materials (“Starbons”) using mesoporous expanded starch as the precursor without the need for a templating agent.

Mesoporous Materials include a range of high surface area porous silicates with applications in gas adsorption, drug delivery, diagnostics and catalysis.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service