Merck
  • Epistatic interactions between neuraminidase mutations facilitated the emergence of the oseltamivir-resistant H1N1 influenza viruses.
Epistatic interactions between neuraminidase mutations facilitated the emergence of the oseltamivir-resistant H1N1 influenza viruses.
Nature communications (2014-10-10)
Susu Duan, Elena A Govorkova, Justin Bahl, Hassan Zaraket, Tatiana Baranovich, Patrick Seiler, Kristi Prevost, Robert G Webster, Richard J Webby
ABSTRACT
Oseltamivir-resistant H1N1 influenza viruses carrying the H275Y neuraminidase mutation predominated worldwide during the 2007-2009 seasons. Although several neuraminidase substitutions were found to be necessary to counteract the adverse effects of H275Y, the order and impact of evolutionary events involved remain elusive. Here we reconstruct H1N1 neuraminidase phylogeny during 1999-2009, estimate the timing and order of crucial amino acid changes and evaluate their impact on the biological outcome of the H275Y mutation. Of the 12 neuraminidase substitutions that occurred during 1999-2009, 5 (chronologically, V234M, R222Q, K329E, D344N, H275Y and D354G) are necessary for maintaining full neuraminidase function in the presence of the H275Y mutation by altering protein accumulation or enzyme affinity/activity. The sequential emergence and cumulative effects of these mutations clearly illustrate a role for epistasis in shaping the emergence and subsequent evolution of a drug-resistant virus population, which can be useful in understanding emergence of novel viral phenotypes of influenza.
Product Description
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
4-Methylumbelliferone, ≥98%
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, suitable for HPLC
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Hymecromone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, ACS reagent