Merck
  • Altered expression of transforming growth factor-beta isoforms in bovine cystic ovarian disease.
Altered expression of transforming growth factor-beta isoforms in bovine cystic ovarian disease.
Reproduction in domestic animals = Zuchthygiene (2014-08-13)
V Matiller, M L Stangaferro, P U Díaz, H H Ortega, F Rey, E Huber, N R Salvetti
ABSTRACT
Cystic ovarian disease (COD) is one of the main causes of infertility in dairy cattle. It has been shown that intra-ovarian factors may contribute to follicular persistence. Transforming growth factor-beta (TGFB) isoforms are important paracrine and autocrine signalling molecules that regulate ovarian follicle growth and physiology. Considering the importance of these factors in the ovarian physiology, in this study, we examined the expression of TGFB isoforms (TGFB1, TGFB2 and TGFB3) in the ovary of healthy cows and animals with spontaneous and adrenocorticotrophic hormone (ACTH)-induced COD. In the oestrous-synchronized control group, the expression of TGFB1 in granulosa and theca cells was higher in spontaneous cysts than in atretic or tertiary follicles. When we compared TGFB2 expression in granulosa cells from atretic or tertiary follicles from the oestrous-synchronized control group with that in ACTH-induced or spontaneous follicular cysts, we found a higher expression in the latter. The expression of the TGFB isoforms studied was also altered during folliculogenesis in both the spontaneous and ACTH-induced COD groups. As it has been previously shown that TGFB influences steroidogenesis, ovarian follicular proliferation and apoptosis, an alteration in its expression may contribute to the pathogenesis of this disease.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%