Skip to Content
Merck
All Photos(2)

Key Documents

450022

Sigma-Aldrich

Sodium fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Sodium monofluoride, Sodium monofluoride (NaF)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050104
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

vapor pressure

1.4 mmHg ( 0 °C)

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

993 °C (lit.)

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium fluoride is a high-purity inorganic compound with a high refractive index and unique luminescent properties. It is widely used in the fields of nanomaterial synthesis, battery materials, organic synthesis, and the dental industry.

Application

Sodium fluoride can be used:
  • As a precursor to synthesize lanthanide fluoride nanoparticles for various applications.
  • To prepare aluminum nitride powder by solution combustion method.
  • As a catalyst for the synthesis of 2,4-disubstituted-1,3-thiazoles andselenazoles, and Friedlander condensation.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced cortical neurotransmitter receptor complex levels in fetal Down syndrome brain.,
Falsafi SK, et al.
Amino Acids, 1-14 (2015)
AlN powder synthesis by sodium fluoride-assisted carbothermal combustion
Haoyang Wu, et al.},
Ceramics International, 40, 14447-14452 (2014)
18F-fluoride for bone imaging
Blau M, et al.
Seminars in Nuclear Medicine, 2(1), 31-37 null
Two-dimensional Acetate-based Light Lanthanide Fluoride Nanomaterials (F?Ln, Ln = La, Ce, Pr, and Nd): Morphology, Structure, Growth Mechanism, and Stability
Leitao Zhang, et al.
Journal of the American Chemical Society, 141, 13134-13142 (2019)
Sodium fluoride as an efficient catalyst for the synthesis of 2,4-disubstituted-1,3-thiazoles and selenazoles at ambient temperature
Janardhan Banothu, et al
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 25, 172-175 (2014)

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service