Skip to Content
Merck
All Photos(3)

Documents

S1504

Sigma-Aldrich

Sodium fluoride

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

1.4 mmHg ( 0 °C)

product line

ReagentPlus®

Assay

≥99%

form

powder

mp

993 °C (lit.)

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Investigations of the reaction mechanism of sodium with hydrogen fluoride to form sodium fluoride and the adsorption of hydrogen fluoride on sodium fluoride monomer and tetramer.: The paper delves into the reaction mechanisms between sodium and hydrogen fluoride, and the adsorption properties of hydrogen fluoride on sodium fluoride structures, which are vital for chemical synthesis and industrial applications (Yu et al., 2024).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa van Kranenburg et al.
Human mutation, 36(2), 200-209 (2014-11-05)
The mechanistic target of rapamycin complex 1 (TORC1) senses nutrient availability to regulate eukaryotic anabolic metabolism. In response to limiting concentrations of amino acids, TORC1 kinase activity is inhibited through the GATOR-1 complex. Mutations in DEPDC5, that encodes one of
Ryan D Espy et al.
Analytical chemistry, 86(15), 7712-7718 (2014-06-28)
Determination of eight drugs of abuse in blood has been performed using paper spray or extraction spray mass spectrometry in under 2 min with minimal sample preparation. A method has been optimized for quantification of amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxy-N-methylamphetamine
Peter Kruzliak et al.
Journal of cellular and molecular medicine, 19(11), 2647-2654 (2015-08-26)
The role of an atherogenic diet in causing pulmonary fibrosis has received little attention and simvastatin has been shown to reduce pulmonary fibrosis in animal models. To determine if an atherogenic diet can induce pulmonary fibrosis and whether simvastatin treatment
E P S Conceição et al.
The Journal of physiology, 593(21), 4799-4811 (2015-09-04)
Neonatal overfeeding induced by litter size reduction leads to further obesity and other metabolic disorders, such as liver oxidative stress and microsteatosis at adulthood. We hypothesized that overfeeding causes an early redox imbalance at weaning, which could programme the animals
M Ogawa et al.
Veterinary pathology, 52(6), 1099-1107 (2015-03-04)
Canine degenerative myelopathy (DM) is a progressive neurodegenerative disease frequently found in Pembroke Welsh Corgi (PWC) dogs, and it has clinical and pathologic similarities to human amyotrophic lateral sclerosis. Autophagy is a major intracellular protein degradation system. Abnormalities of autophagy--resulting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service