Merck
  • FAM83B is a novel biomarker for diagnosis and prognosis of lung squamous cell carcinoma.
FAM83B is a novel biomarker for diagnosis and prognosis of lung squamous cell carcinoma.
International journal of oncology (2015-01-15)
Naoyuki Okabe, Junji Ezaki, Takumi Yamaura, Satoshi Muto, Jun Osugi, Hirosumi Tamura, Jun-Ichi Imai, Emi Ito, Yuka Yanagisawa, Reiko Honma, Mitsukazu Gotoh, Shinya Watanabe, Satoshi Waguri, Hiroyuki Suzuki
ABSTRACT
Personalized therapy for non‑small cell lung cancer (NSCLC), particularly lung adenocarcinoma, has recently been significantly improved by the discovery of various molecular targets. However, this has not been the case for lung squamous cell carcinoma (SCC). In the present study, we identified the family with sequence similarity 83, member B (FAM83B) as a candidate marker for SCC through a comprehensive gene expression analysis and examined its correlations with various clinicopathological factors. The subjects of this study consisted of 215 patients with NSCLC who underwent complete resection from 2005 to 2011 at the Fukushima Medical University Hospital (Fukushima, Japan). They included 102 patients with adenocarcinoma and 113 with SCC. FAM83B expression was first examined in some of the samples by gene expression analysis and western blotting, and then all clinical specimens were evaluated by immunohistochemistry (IHC). The relationship between the quantitative values for IHC and clinicopathological factors was statistically analyzed. The results showed that FAM83B mRNA expression was significantly higher in SCC than in normal lung or adenocarcinoma (P<0.0001). Immunoblot analysis also confirmed this trend. Specimens containing >10% positive area for FAM83B were judged as 'positive'; 94.3% (107/113) of SCC and 14.7% (15/102) of adenocarcinoma were positive. Patients were divided into two subgroups according to expression (54 high‑expression and 53 low‑expression patients); the high‑expression group was associated with a better disease‑free survival (DFS) rate (P=0.042, log‑rank test). In conclusion, FAM83B may be a reliable diagnostic and prognostic biomarker for SCC. Detailed analyses of FAM83B function in lung cancer are required to understand how its expression is associated with better prognosis in SCC.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard