Skip to Content
Merck
All Photos(2)

Documents

05030

Sigma-Aldrich

Sodium dodecyl sulfate solution

greener alternative

BioUltra, for molecular biology, 20% in H2O

Synonym(s):

Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
MDL number:
UNSPSC Code:
12161900
eCl@ss:
39093306
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

description

anionic

product line

BioUltra

form

liquid

mol wt

288.38 g/mol

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

20% in H2O

technique(s)

HPLC: suitable
protein quantification: suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

density

1.03 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
phosphate (PO43-): ≤1 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: ≤0.5
λ: 280 nm Amax: ≤0.3

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

greener alternative category

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is used as a surfactant to enhance catalysis. Click here for more information.

Caution

remark on appearance: may form precipitate at low temperatures, heat to redissolve (15 minutes, 40°C)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Tian et al.
Disease models & mechanisms, 8(6), 635-641 (2015-06-04)
Nemaline myopathy (NM) is one of the most common forms of congenital myopathy, and affects either fast myofibers, slow myofibers, or both. However, an animal model for congenital myopathy with fast-myofiber-specific atrophy is not available. Furthermore, mutations in the leiomodin-3
Lan Zhang et al.
BMC cancer, 14, 885-885 (2014-11-28)
Upregulator of cell proliferation 4 (URG4) has been implicated in the oncogenesis of certain cancers. However, the correlation between URG4 expression and clinicopathological significance in human cancer remains unclear. Therefore, this study investigated its expression and clinicopathological significance in cervical
Jennifer Hirst et al.
Human molecular genetics, 24(17), 4984-4996 (2015-06-19)
Adaptor proteins (AP 1-5) are heterotetrameric complexes that facilitate specialized cargo sorting in vesicular-mediated trafficking. Mutations in AP5Z1, encoding a subunit of the AP-5 complex, have been reported to cause hereditary spastic paraplegia (HSP), although their impact at the cellular
Gerwin Heller et al.
Oncotarget, 6(1), 394-408 (2014-12-17)
In our study, we investigated the role of ZNF677 in non-small cell lung cancers (NSCLC). By comparing ZNF677 expression in primary tumor (TU) and in the majority of cases also of corresponding non-malignant lung tissue (NL) samples from > 1,000
Naoyuki Okabe et al.
International journal of oncology, 46(3), 999-1006 (2015-01-15)
Personalized therapy for non‑small cell lung cancer (NSCLC), particularly lung adenocarcinoma, has recently been significantly improved by the discovery of various molecular targets. However, this has not been the case for lung squamous cell carcinoma (SCC). In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service