Skip to Content
Merck
All Photos(1)

Key Documents

55422

Sigma-Aldrich

Sodium dodecyl sulfate

≥98.0% (GC)

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
Pricing and availability is not currently available.

description

anionic

Quality Level

Assay

≥98.0% (GC)

mol wt

288.38 g/mol

technique(s)

NMR: suitable
ion chromatography: suitable

loss

≤5% loss on drying

mp

204-207 °C (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤200 mg/kg
phosphate (PO43-): ≤1 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Anionic detergent

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shane Stegeman et al.
Cancer discovery, 5(4), 368-379 (2015-02-19)
Prostate cancer is the second most common malignancy among men worldwide. Genome-wide association studies have identified 100 risk variants for prostate cancer, which can explain approximately 33% of the familial risk of the disease. We hypothesized that a comprehensive analysis
A M Maillard et al.
Molecular psychiatry, 20(1), 140-147 (2014-11-26)
Anatomical structures and mechanisms linking genes to neuropsychiatric disorders are not deciphered. Reciprocal copy number variants at the 16p11.2 BP4-BP5 locus offer a unique opportunity to study the intermediate phenotypes in carriers at high risk for autism spectrum disorder (ASD)
Lovisa Lundholm et al.
International journal of radiation biology, 90(8), 718-726 (2014-03-22)
We aimed to analyze the activation status of commonly deregulated receptor tyrosine kinases (RTK) in human non-small cell lung cancer (NSCLC) tumor initiating cells (TIC) previously demonstrated to be refractory to ionizing radiation and chemotherapy. Phosphorylated RTK and important signaling
S Desrivières et al.
Molecular psychiatry, 20(2), 263-274 (2014-02-12)
Despite the recognition that cortical thickness is heritable and correlates with intellectual ability in children and adolescents, the genes contributing to individual differences in these traits remain unknown. We conducted a large-scale association study in 1583 adolescents to identify genes
Kai Wei Kelvin Lee et al.
The ISME journal, 8(4), 894-907 (2013-10-25)
Most studies of biofilm biology have taken a reductionist approach, where single-species biofilms have been extensively investigated. However, biofilms in nature mostly comprise multiple species, where interspecies interactions can shape the development, structure and function of these communities differently from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service