Skip to Content
Merck
All Photos(1)

Key Documents

60218

Sigma-Aldrich

Potassium phosphate monobasic

BioUltra, for molecular biology, anhydrous, ≥99.5% (T)

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

for molecular biology

product line

BioUltra

Assay

≥99.5% (T)

form

powder or crystals

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.001% total nitrogen (N)

pH

4.0-4.5 (25 °C, 1 M in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

2.338 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤30 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.050
λ: 280 nm Amax: 0.045

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium phosphate monobasic and dibasic, are used to create phosphate buffered solutions such as potassium phosphate buffered water and saline solutions used in biological applications.

Other Notes

Component of PBS - in the isolation and analysis of nuclear RNAcitation
Component of PBS - in the isolation and analysis of nuclear RNA[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and analysis of nuclear RNA.
J R Nevins
Methods in enzymology, 152, 234-241 (1987-01-01)
Bruno Buchholz et al.
American journal of physiology. Heart and circulatory physiology, 307(1), H88-H96 (2014-05-06)
Dystrophin is responsible for the mechanical stabilization of the sarcolemma, and it has been shown that it is one of the most sensitive proteins to ischemic injury. However, the enzyme responsible for this proteolysis is still unknown. Isolated rabbit hearts
Angélica Rosa Faria et al.
PLoS neglected tropical diseases, 9(1), e3429-e3429 (2015-01-09)
Visceral leishmaniasis is the most severe form of leishmaniasis. Worldwide, approximately 20% of zoonotic human visceral leishmaniasis is caused by Leishmania infantum, also known as Leishmania chagasi in Latin America. Current diagnostic methods are not accurate enough to identify Leishmania-infected
Stephanie Dille et al.
Infection and immunity, 83(8), 3268-3280 (2015-06-10)
The Chlamydiales are an order of obligate intracellular bacteria sharing a developmental cycle inside a cytosolic vacuole, with very diverse natural hosts, from amoebae to mammals. The clinically most important species is Chlamydia trachomatis. Many uncertainties remain as to how
Casper Skovgaard et al.
Journal of applied physiology (Bethesda, Md. : 1985), 117(10), 1097-1109 (2014-09-06)
The purpose of this study was to examine whether speed endurance training (SET, repeated 30-s sprints) and heavy resistance training (HRT, 80-90% of 1 repetition maximum) performed in succession are compatible and lead to performance improvements in moderately trained endurance

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service