Merck
  • Effects of adding resorbable chitosan microspheres to calcium phosphate cements for bone regeneration.
Effects of adding resorbable chitosan microspheres to calcium phosphate cements for bone regeneration.
Materials science & engineering. C, Materials for biological applications (2014-12-11)
Dan Meng, Limin Dong, Ying Wen, Qiufei Xie
ABSTRACT
Calcium phosphate cements (CPCs) have been widely used as bone graft substitutes. However, the undesirable osteoinductivity and slow degradability of CPCs greatly hamper their clinical application. The aim of this study was to synthesize a type of injectable, bioactive cement. This was accomplished by incorporating chitosan microspheres into CPC. CPC containing chitosan microspheres was analyzed by X-ray diffraction (XRD) and scanning electron microscope (SEM). XRD showed that the hardened chitosan microsphere/CPC with different proportions of microspheres contained diffraction peaks of hydroxyapatite and chitosan. Compressive strength and dissolution in simulated body fluid were measured. The chitosan microsphere/CPC containing 10% (w/w) chitosan microspheres had a compressive strength of 14.78 ± 0.67 MPa. Cavity defects were created in both femoral condylar regions of New Zealand White rabbits. Chitosan microsphere/CPC (composite group) and α-TCP/CPC (control group) were implanted separately into the bone defects of both femurs. X-ray analysis was performed to observe the filling of these bone defects 3 days after surgery. The extent of bone substitute degradation and new bone formation were evaluated by SEM and histological examination at 8, 16, and 24 weeks after implantation. These results showed far more new bone formation and degradation of the chitosan microsphere/CPC composite in the bone defects. These data indicate that a chitosan microsphere/CPC composite might be considered as a promising injectable material for the generation of new bone tissue.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium phosphate monobasic monohydrate, purum p.a., ≥80% (KT)
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%