Skip to Content
Merck
All Photos(1)

Key Documents

71496

Sigma-Aldrich

Sodium phosphate monobasic

purum p.a., anhydrous, ≥99.0% (T)

Synonym(s):

Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4
CAS Number:
Molecular Weight:
119.98
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070208
PubChem Substance ID:
NACRES:
NA.25

grade

anhydrous
purum p.a.

Quality Level

Assay

≥99.0% (T)

form

crystals

pH

4.0-4.5 (25 °C, 50 mg/mL in H2O)

solubility

water: 0.5 g/10 mL, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤300 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

application(s)

sample preparation

SMILES string

[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

AJPJDKMHJJGVTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium phosphate monobasic is an effective oral osmotic laxative at minimal concentration. It is used for colonic cleansing before endoscopic examination of colon.

Application

Sodium phosphate monobasic has been used:
  • as an inorganic phosphate source in cell culture media.
  • in preparation of phosphate buffered saline.
  • in preparation of phosphate buffer in glucose assay.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple microplate-based method for the determination of alpha-amylase activity using the glucose assay kit (GOD method)
Visvanathan R, et al.
Food Chemistry, 211, 853-859 (2016)
Bowel preparation for colonoscopy with sodium phosphate solution versus polyethylene glycol-based lavage: a multicenter trial
Schanz S, et al.
Diagnostic and therapeutic endoscopy., 2008 (2008)
Microcalcifications in breast cancer: novel insights into the molecular mechanism and functional consequence of mammary mineralisation
Cox RF, et al.
British Journal of Cancer, 106(3), 525-525 (2012)
Semiautomated correlative 3D electron microscopy of in vivo-imaged axons and dendrites
Maco B, et al.
Nature Protocols, 9(6), 1354-1354 (2014)
The safety profile of oral sodium phosphate for colonic cleansing before colonoscopy in adults
Hookey LC, et al.
Gastrointestinal Endoscopy, 56(6), 895-902 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service