Skip to Content
Merck
All Photos(3)

Key Documents

481807

Sigma-Aldrich

Calcium carbonate

99.999% trace metals basis

Synonym(s):

Aragonite, Calcite, Calcium monocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
powder
solubility:
H2O: insoluble

Quality Level

Assay

99.999% trace metals basis

form

powder

impurities

≤15.0 ppm Trace Metal Analysis

refractive index

n20/D 1.6584 (lit.)

solubility

H2O: insoluble

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krzysztof Kilian et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-25)
The rapid increase in applications of scandium isotopes in nuclear medicine requires new efficient production routes for these radioisotopes. Recently, irradiations of calcium in cyclotrons by α, deuteron, and proton beams have been used. Therefore, effective post-irradiation separation and preconcentration
Jane A Cauley et al.
Journal of women's health (2002), 22(11), 915-929 (2013-10-18)
Clinical outcomes of the Women's Health Initiative (WHI) calcium plus vitamin D supplementation trial have been reported during 7.0 years of active intervention. We now report outcomes 4.9 years after the intervention stopped and cumulative findings. Postmenopausal women (N=36,282) were
M Azizur Rahman et al.
PloS one, 8(3), e58781-e58781 (2013-03-26)
An organic matrix consisting of a protein-polysaccharide complex is generally accepted as an important medium for the calcification process. While the role this "calcified organic matrix" plays in the calcification process has long been appreciated, the complex mixture of proteins
Christopher T Parsons et al.
Environmental pollution (Barking, Essex : 1987), 178, 254-263 (2013-04-17)
Arsenic contamination of floodplain soils is extensive and additional fresh arsenic inputs to the pedosphere from human activities are ongoing. We investigate the cumulative effects of repetitive soil redox cycles, which occur naturally during flooding and draining, on a calcareous
Pedram Fatehi et al.
Carbohydrate polymers, 94(1), 531-538 (2013-04-03)
In this work, we aimed at adsorbing the oligo-sugars of prehydrolysis liquor on precipitated calcium carbonate (PCC) to produce modified PCC. The results showed that the adsorptions of oligo-sugars, lignin and furfural were greater on porous PCC (PCC2) than on

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service