Merck
  • Foliar application of Zn at flowering stage improves plant's performance, yield and yield attributes of black gram.
Foliar application of Zn at flowering stage improves plant's performance, yield and yield attributes of black gram.
Indian journal of experimental biology (2013-08-01)
Nalini Pandey, Bhavana Gupta, Girish Chandra Pathak
ABSTRACT
Black gram plants subjected to varying levels of Zn supply (0.01 to 10 microM Zn) showed optimum growth and dry matter yield in plants receiving 1 microM Zn. The dry matter yield of plants decreased in plants receiving 0.01 and 0.1 microM Zn (deficient) and excess levels of Zn (2 and 10 microM Zn). The plants grown with Zn deficient supply showed delayed flowering, premature bud abscission, reduced size of anthers, pollen producing capacity, pollen viability and stigma receptivity resulting in poor pod formation and seed yield. Providing Zn as a foliar spray at pre-flowering stage minimized the severity of Zn deficiency on reproductive structure development and enhanced the seed nutritional status by enhancing seed Zn density, seed carbohydrate (sugar and starch content) and storage proteins (albumins, globulins, glutenins, and prolamines).
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis