Merck
  • Physical entrapment of hyaluronic acid during synthesis results in extended release from model hydrogel and silicone hydrogel contact lens materials.
Physical entrapment of hyaluronic acid during synthesis results in extended release from model hydrogel and silicone hydrogel contact lens materials.
Eye & contact lens (2013-02-16)
Andrea Weeks, Lakshman N Subbaraman, Lyndon Jones, Heather Sheardown
ABSTRACT
This study was designed to assess the duration of hyaluronic acid (HA) release from model contact lens materials when HA was physically incorporated into the hydrogel during synthesis and to assess the effects of the HA release on lysozyme sorption. Model conventional and silicone hydrogel contact lens materials containing HA of various molecular weights as a releasable wetting agent were prepared. The HA was released into phosphate-buffered saline and MilliQ water, and the release was monitored using ultraviolet spectroscopy. Hyaluronic acid release was quantified by enzyme-linked immunosorbent assay. The effect of the releasable HA on lysozyme sorption to the materials was also analyzed using 125-I-labeled protein. Hyaluronic acid loaded into the materials using this method could be released from conventional hydrogel materials for 21 days; the model silicone hydrogels showed release of more than 7 weeks. With one exception, the releasable HA decreased lysozyme sorption. Hyaluronic acid physically incorporated into contact lens materials during synthesis may therefore be released for extended periods of time of up to 7 weeks. Hyaluronic acid release leads to decreased protein adsorption in general. This method has potential for modification of conventional and silicone hydrogel lenses with releasable HA as a wetting agent.
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, ≥100,000 units/mg protein, lyophilized powder
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein