Skip to Content
Merck
All Photos(1)

Key Documents

1356676

USP

Anhydrous lactose

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Lactose, (+)-Lactose, 4-O-Β-D-Galactopyranosyl-D-glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93796
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

lactose

manufacturer/tradename

USP

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h1,4-12,14-21H,2-3H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-/m0/s1

InChI key

DKXNBNKWCZZMJT-JVCRWLNRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Anhydrous lactose USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Lactulose Concentrate
  • Lactulose Solution
  • Galactose
  • Compressible Sugar
  • Sugar Spheres
  • Confectioner′s Sugar

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lactulose Concentrate
United States Pharmacopeia, 40(5), 2521-2521 (2016)
Galactose
United States Pharmacopeia, 44(5) (2020)
Lactulose Solution
United States Pharmacopeia, 40(5, 2522-2522 (2016)
Anhydrous Lactose
United States Pharmacopeia, 46(5), 5847-5847 (2020)
Pedro M R Guimarães et al.
Biotechnology advances, 28(3), 375-384 (2010-02-16)
Cheese whey, the main dairy by-product, is increasingly recognized as a source of many bioactive valuable compounds. Nevertheless, the most abundant component in whey is lactose (ca. 5% w/v), which represents a significant environmental problem. Due to the large lactose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service