Skip to Content
Merck
All Photos(1)

Documents

61345

Sigma-Aldrich

D-Lactose monohydrate

≥98.0% (HPLC)

Synonym(s):

β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Milk sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
3768231
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥98.0% (HPLC)

form

solid

optical activity

[α]20/D +52±2°, 22 hr, c = 10% in H2O

impurities

3.0-7.0% water

color

colorless

solubility

H2O: 0.1 g/mL, clear, colorless (warm)

SMILES string

O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1

InChI key

HBDJFVFTHLOSDW-XBLONOLSSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis of lactose lauryl ester in organic solvents using aluminosilicate zeolite as a catalyst.: This research explores the synthesis of lactose lauryl ester, utilizing D-lactose monohydrate and organic solvents in the presence of aluminosilicate zeolite as a catalyst, highlighting its potential applications in food chemistry and surfactants (Enayati et al., 2019).

  • Synthesis and characterization of lactose fatty acid ester biosurfactants using free and immobilized lipases in organic solvents.: The paper presents the synthesis and characterization of lactose fatty acid esters using D-lactose monohydrate and various lipases, focusing on their potential as biosurfactants in food and cosmetic industries (Enayati et al., 2018).

Biochem/physiol Actions

Lactose is a dissacharide formed by the condensation of one galactose and one glucose molecule. Lactose is the major sugar in the milk of most species, typically present between 2-8%. The enzyme lactase hydrolyzes lactose to its constituent monosaccharides. In neonates, glucose released via the action of lactase is a major energy source.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mojtaba Enayati et al.
Food chemistry, 279, 401-407 (2019-01-07)
Sugar-based biosurfactants are safer and healthier alternatives to synthetic surfactants especially for use in the food industry. In this work, biosurfactants were synthesized via the esterification of lactose with lauric acid in different organic solvents without using lipase or other
Erica S Rodriguez et al.
Journal of the science of food and agriculture, 99(4), 1682-1690 (2018-09-13)
Chia oil possesses a very high content of polyunsaturated fatty acids, mainly α-linolenic acid. This characteristic makes this oil possess beneficial properties to health but gives it a high susceptibility to the oxidation process. Microencapsulation and the addition of natural
Jiseon Yang et al.
NPJ biofilms and microbiomes, 7(1), 70-70 (2021-09-08)
While sequencing technologies have revolutionized our knowledge of microbial diversity, little is known about the dynamic emergent phenotypes that arise within the context of mixed-species populations, which are not fully predicted using sequencing technologies alone. The International Space Station (ISS)
Maryam Enteshari et al.
Foods (Basel, Switzerland), 9(6) (2020-06-18)
The simultaneous production of lactulose (LAU), lactobionic acid (LBA), and organic acids from sweet and acid whey permeate (SWP and AWP) via catalytic synthesis (5% Ru/C) was studied in a continuous stirred-tank reactor. At selected conditions (60 °C, 60 bar
Fan Zhou et al.
Scientific reports, 1, 78-78 (2012-02-23)
The invisibility cloak has been a long-standing dream for many researchers over the decades. Using transformation optics, a three-dimensional (3D) object is perceived as having a reduced number of dimensions, making it "undetectable" judging from the scattered field12345. Despite successful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service