Lactose is a dissacharide formed by the condensation of one galactose and one glucose molecule. Lactose is the major sugar in the milk of most species, typically present between 2-8%. The enzyme lactase hydrolyzes lactose to its constituent monosaccharides. In neonates, glucose released via the action of lactase is a major energy source.
Other Notes
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Colloids and surfaces. B, Biointerfaces, 123, 53-60 (2014-09-30)
Segregation of the protein bovine serum albumin (BSA) and lactose in thin aqueous films during drying was investigated by examining the composition of the dried films using inverse micro Raman spectroscopy (IMRS) and X-ray photoelectron spectroscopy (XPS) sputter-depth profiling. The
The invisibility cloak has been a long-standing dream for many researchers over the decades. Using transformation optics, a three-dimensional (3D) object is perceived as having a reduced number of dimensions, making it "undetectable" judging from the scattered field12345. Despite successful
Gaucher disease type 1 is characterized by hepatosplenomegaly, anemia, thrombocytopenia, and skeletal disease. A safe, effective oral therapy is needed. To determine whether eliglustat, a novel oral substrate reduction therapy, safely reverses clinical manifestations in untreated adults with Gaucher disease
Journal of pharmaceutical and biomedical analysis, 107, 318-324 (2015-02-01)
This paper reports the application of surface enhanced Raman chemical imaging (SER-CI) as a potentially non-destructive quantitative analytical method for the investigation of model pharmaceutical formulations containing the active pharmaceutical ingredient (API) in low concentrations (0.5-2%). The application of chemometric
Chemical & pharmaceutical bulletin, 63(2), 95-101 (2015-03-10)
The manufacture of highly drug-loaded fine globular granules eventually applied for orally disintegrating tablets has been investigated using a unique multi-functional rotor processor with acetaminophen, which was used as a model drug substance. Experimental design and statistical analysis were used
This product is not sterile. A sterile product is not available at this time.
Helpful?
Reviews
★★★★★ No rating value