Skip to Content
Merck
All Photos(1)

Key Documents

1200000

USP

Digoxin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

12β-Hydroxydigitoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H64O14
CAS Number:
Molecular Weight:
780.94
Beilstein:
77011
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

digoxin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]2[C@@H](O)C[C@@H](O[C@@H]2C)O[C@H]3[C@@H](O)C[C@@H](O[C@@H]3C)O[C@H]4CC[C@@]5(C)[C@H](CC[C@@H]6[C@@H]5C[C@@H](O)[C@]7(C)[C@H](CC[C@]67O)C8=CC(=O)OC8)C4

InChI

1S/C41H64O14/c1-19-36(47)28(42)15-34(50-19)54-38-21(3)52-35(17-30(38)44)55-37-20(2)51-33(16-29(37)43)53-24-8-10-39(4)23(13-24)6-7-26-27(39)14-31(45)40(5)25(9-11-41(26,40)48)22-12-32(46)49-18-22/h12,19-21,23-31,33-38,42-45,47-48H,6-11,13-18H2,1-5H3/t19-,20-,21-,23-,24+,25-,26-,27+,28+,29+,30+,31-,33+,34+,35+,36-,37-,38-,39+,40+,41+/m1/s1

InChI key

LTMHDMANZUZIPE-PUGKRICDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Digoxin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Digoxin Tablets
  • Digoxin Injection
  • Digoxin Oral Solution

Biochem/physiol Actions

A cardiac glycoside, substrate for Pgp, upregulates Pgp expression and down regulates SXR (Steroid Xenobiotic Receptor).
A cardiac glycoside, substrate for Pgp.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert J DiDomenico et al.
Pharmacotherapy, 34(11), 1121-1131 (2014-08-29)
To compare the frequency of achieving a therapeutic serum digoxin concentration (SDC), defined as 0.5-0.9 ng/ml, by using a simplified nomogram to individualize digoxin dosing with standard dosing practices in patients with heart failure, and to characterize the relationship between
Susanne Johansson et al.
Clinical pharmacokinetics, 53(9), 837-847 (2014-08-15)
Vandetanib is a selective inhibitor of vascular endothelial growth factor receptor (VEGFR), epidermal growth factor receptor (EGFR) and rearranged during transfection (RET) signalling, indicated for the treatment of medullary thyroid cancer. We investigated potential drug-drug interactions between vandetanib and metformin
Mayank K Mittal et al.
American journal of therapeutics, 18(4), 280-287 (2010-03-13)
Heart failure (HF) is a leading cause of morbidity and mortality. Appropriate medical therapy using angiotensin converting enzyme inhibitors and beta-blockers improves outcomes in HF, whereas the role of digoxin is still not clearly defined. Digoxin is currently recommended for
Emma Eade et al.
International journal of cardiology, 164(3), 365-367 (2011-08-03)
Digoxin has been used for hundreds of years to aid rate control in atrial fibrillation and as a positive inotrope in heart failure. Our familiarity with digoxin has allowed it to become easily prescribed even though there are data suggesting
Chang Gong et al.
Cancer research, 74(16), 4341-4352 (2014-07-02)
Phyllodes tumors of breast, even histologically diagnosed as benign, can recur locally and have metastatic potential. Histologic markers only have limited value in predicting the clinical behavior of phyllodes tumors. It remains unknown what drives the malignant progression of phyllodes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service