Skip to Content
Merck
All Photos(1)

Documents

1199002

USP

Digitoxin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5β,20(22)-Cardenolide-3β,14-diol-3-(O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy, Digitoxoside, Lanatoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H64O13
CAS Number:
Molecular Weight:
764.94
Beilstein:
76678
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

digitoxin

manufacturer/tradename

USP

mp

240 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]1(C[C@H](O)[C@H](O[C@@]2([H])C[C@H](O)[C@H](O[C@@]3([H])C[C@H](O)[C@H](O)[C@@H](C)O3)[C@@H](C)O2)[C@@H](C)O1)O[C@H]4CC[C@@]5(C)[C@]([H])(CC[C@]6([H])[C@]5([H])CC[C@]7(C)[C@]([H])(CC[C@]67O)C8=CC(=O)OC8)C4

InChI

1S/C41H64O13/c1-20-36(46)29(42)16-34(49-20)53-38-22(3)51-35(18-31(38)44)54-37-21(2)50-33(17-30(37)43)52-25-8-11-39(4)24(15-25)6-7-28-27(39)9-12-40(5)26(10-13-41(28,40)47)23-14-32(45)48-19-23/h14,20-22,24-31,33-38,42-44,46-47H,6-13,15-19H2,1-5H3/t20-,21-,22-,24-,25+,26-,27+,28-,29+,30+,31+,33+,34+,35+,36-,37-,38-,39+,40-,41+/m1/s1

InChI key

WDJUZGPOPHTGOT-XUDUSOBPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Digitoxin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Digitoxin Injection
  • Digitoxin Tablets
  • Digoxin

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

[Digitoxin].
K Yamamoto et al.
Nihon rinsho. Japanese journal of clinical medicine, 53 Su Pt 1, 945-949 (1995-02-01)
Miguel López-Lázaro
Expert opinion on therapeutic targets, 11(8), 1043-1053 (2007-08-02)
Accumulating preclinical and clinical data suggest that the cardiac drug digitoxin might be used in cancer therapy. Recent reports have shown that digitoxin can inhibit the growth and induce apoptosis in cancer cells at concentrations commonly found in the plasma
[Digitoxin overdose with reversible thrombocytopenia].
K G Dahmen et al.
Medizinische Klinik (Munich, Germany : 1983), 91(12), 791-795 (1996-12-15)
G G Belz et al.
European journal of clinical investigation, 31 Suppl 2, 10-17 (2001-08-30)
Digitalis glycosides exert a positive inotropic effect, i.e. an increase in myocardial contractility associated with a prolongation of relaxation period, and glycosides lower the heart rate (negative chronotropic), impede stimulus conduction (negative dromotropic) and promote myocardial excitability (positive bathmotropic). They
H F Vöhringer et al.
International journal of clinical pharmacology, therapy, and toxicology, 19(4), 175-184 (1981-04-01)
When prescribing cardiac glycosides for patients with renal failure, one should consider the different pharmacokinetics of the two most important glycosides, digoxin and digitoxin. Whereas steady state plasma concentrations of digoxin are altered proportionally to renal clearance of creatinine, those

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service